EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Augmentation of NAD(+) by NQO1 attenuates cisplatin-mediated hearing impairment.
PMID 24922076
Abstract
Cisplatin (cis-diaminedichloroplatinum-II) is an extensively used chemotherapeutic agent, and one of its most adverse effects is ototoxicity. A number of studies have demonstrated that these effects are related to oxidative stress and DNA damage. However, the precise mechanism underlying cisplatin-associated ototoxicity is still unclear. The cofactor nicotinamide adenine dinucleotide (NAD(+)) has emerged as a key regulator of cellular energy metabolism and homeostasis. Here, we demonstrate for the first time that, in cisplatin-mediated ototoxicity, the levels and activities of SIRT1 are suppressed by the reduction of intracellular NAD(+) levels. We provide evidence that the decrease in SIRT1 activity and expression facilitated by increasing poly(ADP-ribose) transferase (PARP)-1 activation and microRNA-34a through p53 activation aggravates cisplatin-mediated ototoxicity. Moreover, we show that the induction of cellular NAD(+) levels using β-lapachone (β-Lap), whose intracellular target is NQO1, prevents the toxic effects of cisplatin through the regulation of PARP-1 and SIRT1 activity. These results suggest that direct modulation of cellular NAD(+) levels by pharmacological agents could be a promising therapeutic approach for protection from cisplatin-induced ototoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1390
3,3′-Methylene-bis(4-hydroxycoumarin)
C19H12O6
SAB4501989
Anti-NF-κB p105 antibody produced in rabbit, affinity isolated antibody
SAB4300301
Anti-NFKB1 (Ab-337) antibody produced in rabbit, affinity isolated antibody
SAB4300302
Anti-NFKB1 (Ab-893) antibody produced in rabbit, affinity isolated antibody
SAB4300303
Anti-NFKB1 (Ab-907) antibody produced in rabbit, affinity isolated antibody
SAB4300554
Anti-NFKB1 (Ab-927) antibody produced in rabbit, affinity isolated antibody
SAB4300498
Anti-NFKB1 (Ab-932) antibody produced in rabbit, affinity isolated antibody
GW21167 Anti-NFKB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406182
Anti-NFKB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38413
Anti-NFKB1 antibody produced in rabbit, affinity isolated antibody
HPA027305
Anti-NFKB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103906
Anti-NFKB1 antibody produced in rabbit, affinity isolated antibody
N8523
Monoclonal Anti-NF-κB antibody produced in mouse, clone NF-12, ascites fluid
WH0004790M1
Monoclonal Anti-NFKB1 antibody produced in mouse, clone 2E6, purified immunoglobulin, buffered aqueous solution
PLA0214
Rabbit anti-NF-kappaB1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.