EMAIL THIS PAGE TO A FRIEND
Anticancer research
Cell-adhesion molecule expression and the proliferation of malignant mesothelioma: a post-mortem examination.
PMID 24922673
Abstract
In order to determine if metastatic malignant mesothelioma cells are more aggressive than primary malignant mesothelioma cells, an analysis of the expression of the adhesion molecules E-cadherin and β-catenin, concomitant with an assessment of the proliferative activity at primary and metastatic sites, was conducted in post-mortem samples. E-cadherin or β-catenin expression was graded according to the percentage of positively-stained tumor cells. The proliferative activity was quantified by the Ki-67 labeling index. Histologically, the majority of metastatic tumors matched the primary tumor. In the epithelioid component of primary tumors, E-cadherin and β-catenin expression ranged from 1+ to 4+. Malignant mesothelioma cells acquire a higher proliferative potential after metastasis, without any significant changes in their histology, although metastasis produces no definite trend on the expression of E-cadherin or β-catenin.