EMAIL THIS PAGE TO A FRIEND
Fitoterapia
New cholinesterase inhibitors from Garcinia atroviridis.
PMID 24924287
Abstract
A triflavanone, Garcineflavanone A (1) and a biflavonol, Garcineflavonol A (2) have been isolated from the stem bark of Garcinia atroviridis (Clusiaceae), collected in Peninsular Malaysia. Their structures were established using one and two-dimensional NMR, UV, IR and mass spectrometry and evaluated in vitro for their acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) enzymes inhibitory activity. Molecular docking studies of the isolated compounds were performed using docking procedure of AutoDock to disclose the binding interaction and orientation of these molecules into the active site gorge.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8375
Eserine
C15H21N3O2
45710
Eserine, ≥98.0% (N)
C15H21N3O2
B3128
S-Butyrylthiocholine chloride, crystalline
C9H20ClNOS