EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Loss of Nrf2 in mice evokes a congenital intrahepatic shunt that alters hepatic oxygen and protein expression gradients and toxicity.
PMID 24924401
Abstract
The transcription factor Nrf2 (Nfe2l2 nuclear factor, erythroid 2-like 2) regulates gene expression directly, controlling pharmacological and toxicological responses. These processes may also be influenced by the structure of the hepatic vasculature, which distributes blood flow through compartmentalized microenvironments to maintain organismal stability. Castings of the hepatic portal vasculature of albino C57BL/6J but not ICR Nrf2(-/-) mice revealed a congenital intrahepatic shunt that was present in two thirds of Nrf2-disrupted mice. This shunt directly connected the portal vein to the inferior vena cava and displayed characteristics of a patent ductus venosus. Immunohistochemistry revealed that Nrf2(-/-) mice with an intrahepatic shunt manifest changes to hepatic oxygen and protein expression gradients when compared with wild-type (WT) and non-shunted Nrf2(-/-) mice. Centrilobular hypoxia found in WT and Nrf2(-/-) mice without shunts was reduced in Nrf2(-/-) livers with a shunt. Hepatic protein expression of phosphoenolpyruvate carboxykinase (Pepck), normally confined to the periportal zone, exhibited both periportal and centrilobular zonal expression in livers from Nrf2(-/-) mice with an intrahepatic shunt. Centrilobular expression of Cytochrome P450 2E1 (Cyp2e1) was diminished in shunted Nrf2(-/-) livers compared with WT and Nrf2(-/-) livers without shunts. The intrahepatic shunt in Nrf2(-/-) mice was further found to diminish acetaminophen hepatoxicity compared with WT and Nrf2(-/-) non-shunted mice following a 6 h challenge with 250 mg/kg acetaminophen. The presence of an intrahepatic shunt influences several physiological and pathophysiological properties of Nrf2(-/-) mice through changes in blood flow, hepatic oxygenation, and protein expression that extent beyond loss of canonical transactivation of Nrf2 target genes.
Related Materials