EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Determination of irinotecan and its metabolite SN-38 in rabbit plasma and tumors using a validated method of tandem mass spectrometry coupled with liquid chromatography.
PMID 24927278
Abstract
New tandem mass spectrometric method coupled with liquid chromatography (LC-MS/MS) has been developed to determine the total concentration of camptothecin derivatives (irinotecan and SN-38) regardless of inter-conversion phenomenon between carboxylate and lactone forms. At first, all sample solutions were acidified for 1h in order to completely convert CPT derivatives into their lactone forms and then CPT derivatives were extracted with organic solution containing diethyl ether and ethyl acetate (2:1, v/v) just after alkalization in the range pH 8.0-8.5 in acid-treated solutions. Analytes were separated on a reverse phase C18 column (150×2.1mm) and eluted isocratically with a mobile phase which consisted of acetonitrile-methanol-buffer (0.1% formic acid, 5mM ammonium formate) (3:4:3, v/v). CPT derivatives were monitored by tandem mass spectrometry in electrospay-positive ionization and multiple reaction mode programmed to the following transitions (m/z): '587.6→167.2' of CPT-11, '393.6→349.3' of SN-38 and '349.4→ 305.2' of CPT. The method was validated to have the proper linearity (r(2)>0.99) over the range of 5-1000ng/ml of CPT-11 and 1-250ng/ml of SN-38 with good accuracy (89.8-114.3%) and precision (less than 10%). In all stability tests, concentration of CPT-11 and SN-38 had been left in the acceptable range of 88.8-110.7% when sample solutions were acidified before determination of CPT derivatives. Newly developed LC-MS/MS method was suitable for the determination of CPT derivatives of both rabbit plasma and tumor tissues in the pharmacokinetic study.