EMAIL THIS PAGE TO A FRIEND
BMC cancer
Tyrosine phosphorylation profiling via in situ proximity ligation assay.
PMID 24928687
Abstract
Tyrosine phosphorylation (pTyr) is an important cancer relevant posttranslational modification since it regulates protein activity and cellular localization. By controlling cell growth and differentiation it plays an important role in tumor development. This paper describes a novel approach for detection and visualization of a panel of pTyr proteins in tumors using in situ proximity ligation assay. K562 leukemia cells were treated with tyrosine kinase and/or phosphatase inhibitors to induce differences in pTyr levels and mimic cells with different malignant properties. Cells were then probed with one antibody against the pTyr modification and another probe against the detected protein, resulting in a detectable fluorescent signal once the probes were in proximity. Total and protein specific pTyr levels on ABL, SHC, ERK2 and PI3K proteins were detected and samples of control and treated cells were distinguished at the pTyr level using this novel approach. Promising results were also detected for formalin fixed and paraffin embedded cells in the micro array format. This application of in situ proximity ligation assay is valuable in order to study the pTyr modification of a panel of proteins in large data sets to validate mass spectrometric data and to be combined with tissue microarrays. The approach offers new opportunities to reveal the pTyr signatures in cells of different malignant properties that can be used as biomarker of disease in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-333
Anti-MAP Kinase 2/Erk2 Antibody, Anti-MAP Kinase 2/Erk2 Antibody detects level of MAP Kinase 2/Erk2 & has been published & validated for use in IP & WB.
GW22768 Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA030069
Anti-MAPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101801
Anti-PI3 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101802
Anti-PI3 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1409446
Anti-PI3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA017737
Anti-PI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305558 MONOCLONAL ANTI-ERK2 antibody produced in mouse, clone 814CT5.4.2, IgG fraction of antiserum, buffered aqueous solution
WH0005594M1
Monoclonal Anti-MAPK1 antibody produced in mouse, clone 1D1, purified immunoglobulin, buffered aqueous solution
WH0005266M1
Monoclonal Anti-PI3 antibody produced in mouse, clone 3G9, purified immunoglobulin, buffered aqueous solution
SAB1409445
Monoclonal Anti-PI3 antibody produced in mouse, clone 2G20, purified immunoglobulin, buffered aqueous solution
PLA0236
Rabbit anti-ERK2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.