EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antimicrobial activity of amine linked bis- and tris-heterocycles.
PMID 24929288
Abstract
A series of amine linked bis- and tris-heterocycles were prepared from heteroaryl cinnamamides and tested for antimicrobial activity. The compounds 11c and 12c exhibited excellent antibacterial activity while 12a and 12c displayed excellent antifungal activity.