EMAIL THIS PAGE TO A FRIEND
Oncogene
Cellular senescence checkpoint function determines differential Notch1-dependent oncogenic and tumor-suppressor activities.
PMID 24931169
Abstract
Notch activity regulates tumor biology in a context-dependent and complex manner. Notch may act as an oncogene or a tumor-suppressor gene even within the same tumor type. Recently, Notch signaling has been implicated in cellular senescence. Yet, it remains unclear as to how cellular senescence checkpoint functions may interact with Notch-mediated oncogenic and tumor-suppressor activities. Herein, we used genetically engineered human esophageal keratinocytes and esophageal squamous cell carcinoma cells to delineate the functional consequences of Notch activation and inhibition along with pharmacological intervention and RNA interference experiments. When expressed in a tetracycline-inducible manner, the ectopically expressed activated form of Notch1 (ICN1) displayed oncogene-like characteristics inducing cellular senescence corroborated by the induction of G0/G1 cell-cycle arrest, Rb dephosphorylation, flat and enlarged cell morphology and senescence-associated β-galactosidase activity. Notch-induced senescence involves canonical CSL/RBPJ-dependent transcriptional activity and the p16(INK4A)-Rb pathway. Loss of p16(INK4A) or the presence of human papilloma virus (HPV) E6/E7 oncogene products not only prevented ICN1 from inducing senescence but permitted ICN1 to facilitate anchorage-independent colony formation and xenograft tumor growth with increased cell proliferation and reduced squamous-cell differentiation. Moreover, Notch1 appears to mediate replicative senescence as well as transforming growth factor-β-induced cellular senescence in non-transformed cells and that HPV E6/E7 targets Notch1 for inactivation to prevent senescence, revealing a tumor-suppressor attribute of endogenous Notch1. In aggregate, cellular senescence checkpoint functions may influence dichotomous Notch activities in the neoplastic context.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405602
Anti-CDKN2B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV03047 Anti-CDKN2B antibody produced in rabbit, affinity isolated antibody
SAB4501594
Anti-Involucrin, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV48675
Anti-IVL antibody produced in rabbit, affinity isolated antibody
HPA055211
Anti-IVL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21943 Anti-MRPL28 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407017
Anti-MRPL28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA030594
Anti-MRPL28 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105816
Anti-MRPL28 antibody produced in rabbit, affinity isolated antibody
SAB4502019
Anti-NOTCH1 (Cleaved-Val1754) antibody produced in rabbit, affinity isolated antibody
N4788 Anti-NOTCH1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101618
Anti-NOTCH1 antibody produced in rabbit, affinity isolated antibody
SAB2501781
Anti-NOTCH3 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
N5038 Anti-NOTCH3 antibody produced in rabbit, affinity isolated antibody
HPA044392
Anti-NOTCH3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306675
ANTI-P53 (T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503015
Anti-p53 antibody produced in rabbit, affinity isolated antibody
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
SAB1306006
ANTI-P53(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503001
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503003
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503010
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503011
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503012
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503013
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503019
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503021
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody