EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Lactose electroisomerization into lactulose: effect of the electrode material, active membrane surface area-to-electrode surface area ratio, and interelectrode-membrane distance.
PMID 24931526
Abstract
The aim of the present work was to study and develop an innovative, clean, and environmentally friendly process for lactulose synthesis by electroactivation of lactose. In this work, the electrode material (type 304 stainless steel, titanium, and copper), dimensionless interelectrode-membrane distance at the cathodic compartment (0.36, 0.68, and 1), and the membrane:electrode surface area ratio (0.23, 0.06, and 0.015) were considered to be the factors that could affect the kinetic conversion of lactose into lactulose. The reactions were conducted under an initial lactose concentration of 0.15mol/L at 10°C, Froude number (mixing speed) of 2.05×10(-2), and electric current intensity of 300mA for 30min. The highest lactulose formation yield of 32.50% (0.05mol/L) was obtained by using a copper electrode, interelectrode-membrane distance of 0.36, and membrane:electrode surface area ratio of 0.23. The 2-parameter Langmuir, Freundlich, and Temkin isotherm models were used for the prediction of the lactose isomerization kinetics as well as the 3-parameter Langmuir-Freundlich isotherm model. It was shown that the lactose isomerization kinetics into lactulose followed the Temkin and Langmuir-Freundlich models with coefficients of determination of 0.99 and 0.90 and a relative error of 1.42 to 1.56% and 4.27 to 4.37%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
202940
Calcium chloride hydrate, 99.999% trace metals basis
CaCl2 · xH2O
48260
D-(+)-Galactose, for microbiology, ≥99.0%
C6H12O6
61339
D-Lactose monohydrate, BioUltra, ≥99.5% (HPLC)
C12H22O11 · H2O
61345
D-Lactose monohydrate, ≥98.0% (HPLC)
C12H22O11 · H2O
61341
D-Lactose monohydrate, tested according to Ph.Eur.
C12H22O11 · H2O
L254
D-Lactose monohydrate, ACS reagent
C12H22O11 · H2O
F2793
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
C6H12O6
47740
D-(−)-Fructose, for microbiology, ≥99.0%
C6H12O6
47748
D-(−)-Fructose, tested according to Ph.Eur.
C6H12O6
1286504
Fructose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
F0550000
Fructose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
PHR1002
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
1287700 Galactose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
PHR1206
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
G0050000
Galactose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
PHR1024
Lactose (Monohydrate), pharmaceutical secondary standard; traceable to USP and PhEur
C12H22O11 · H2O
1356701
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
C12H22O11 · H2O
L7877
Lactulose, ≥95%
C12H22O11
1356803
Lactulose, United States Pharmacopeia (USP) Reference Standard
C12H22O11
L0130000
Lactulose, European Pharmacopoeia (EP) Reference Standard
C12H22O11
61360
Lactulose, ≥98.0% (HPLC)
C12H22O11
PHR1608
Lactulose, Pharmaceutical Secondary Standard; Certified Reference Material
C12H22O11