EMAIL THIS PAGE TO A FRIEND
Oncology letters
Capillary morphogenesis gene 2 regulates adhesion and invasiveness of prostate cancer cells.
PMID 24932305
Abstract
Capillary morphogenesis gene 2 (CMG2), also known as anthrax toxin receptor 2, has been indicated in the formation of new vasculature and in the internalisation of the anthrax toxin. Anti-angiogenesis therapy that targets this molecule has been investigated. However, our recent studies of this molecule have indicated that this gene may also play certain roles in cancer cells. The present study aimed to examine the expression of CMG2 in prostate cancer tissues and cell lines, and also its impact on cellular functions. The expression of CMG2 was detectable in normal and prostate cancer tissues. The prostate cancer cell lines appeared to have relatively high expression compared with the prostatic epithelial cells. Knockdown of CMG2 impaired the adherence of the prostate cancer cells. CMG2 overexpression resulted in decreasing invasiveness, while the knockdown of CMG2 contrastingly enhanced this ability. The altered expression of CMG2 in the prostate cancer cells did not affect the in vitro or in vivo growth of the cells. Taken together, these results show that CMG2 is expressed in prostatic epithelia and cancer cells. In addition to its role in the angiogenesis and the internalisation of anthrax toxin, CMG2 also plays an important role in regulating the adhesion and invasion of prostate cancer cells.