EMAIL THIS PAGE TO A FRIEND
Food & function
The effects of daily supplementation of Dendrobium huoshanense polysaccharide on ethanol-induced subacute liver injury in mice by proteomic analysis.
PMID 24933018
Abstract
Polysaccharides isolated from edible Dendrobium huoshanense have been shown to possess a hepatoprotection function for selenium- and carbon tetrachloride-induced liver injury. In this study, we investigated the preventive effects of daily supplementation with an homogeneous polysaccharide (DHP) purified from D. huoshanense on ethanol-induced subacute liver injury in mice and its potential mechanisms in liver protection by a proteomic approach. DHP was found to effectively depress the increased ratio of liver weight to body weight, reduce the elevated levels of serum aspartate aminotransferase, total cholesterol, total bilirubin and low density lipoprotein, and alleviate hepatic steatosis in mice with ethanol-induced subacute liver injury. Hepatic proteomics analysis performed by two-dimensional difference gel electrophoresis (2D-DIGE) coupled with matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF/TOF-MS) revealed that cystathionine beta-synthase (Cbs) and D-lactate dehydrogenase (Ldhd) were two key proteins regulated by daily DHP intervention, which may assist in correcting the abnormal hepatic methionine metabolism pathway and decreasing the level of hepatic methylglyoxal generated from disordered metabolic pathways caused by ethanol. Our data suggest that DHP can protect liver function from alcoholic injury with complicated molecular mechanisms involving regulation of Cbs and Ldhd.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405568
Anti-CBS antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001223
Anti-CBS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV45746
Anti-CBS antibody produced in rabbit, IgG fraction of antiserum
SAB1411562
Anti-CBS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1408426
Anti-LDHD antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2103557
Anti-LDHD antibody produced in rabbit, affinity isolated antibody
HPA048639 Anti-LDHD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA041766
Anti-LDHD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410326
Anti-LDHD antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV41398
Anti-MAT1A (AB1) antibody produced in rabbit, IgG fraction of antiserum
HPA048627
Anti-MAT1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000875M1
Monoclonal Anti-CBS antibody produced in mouse, clone 3E1, purified immunoglobulin, buffered aqueous solution
SAB1409946
Monoclonal Anti-CBS antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution
SAB1403642
Monoclonal Anti-CBS, (N-terminal) antibody produced in mouse, clone 6B8, purified immunoglobulin, buffered aqueous solution
SAB1402264
Monoclonal Anti-MAT1A antibody produced in mouse, clone 4D11, purified immunoglobulin, buffered aqueous solution