EMAIL THIS PAGE TO A FRIEND
Clinical and experimental medicine
Association of circulating levels of RANTES and -403G/A promoter polymorphism to acute heart failure after STEMI and to cardiogenic shock.
PMID 24934326
Abstract
Chemokines, including RANTES, play a crucial role in the processes of inflammation during cardiovascular disorders, including myocardial infarction, disease progression and complications. This study aimed to evaluate the role of RANTES -403G/A polymorphism and levels in circulation in processes of development and progression of myocardial infarction and cardiogenic shock. A total of 609 patients with ST-segment elevation myocardial infarction, 43 patients with cardiogenic shock and 130 control subjects were enrolled in the study. RANTES -403G/A promoter polymorphism and baseline serum RANTES levels were analyzed. In the present study, we associated RANTES -403G/A promoter polymorphism with acute heart failure in patients with myocardial infarction (pxa0=xa00.006) and ejection fraction 3xa0months after MI onset (pxa0=xa00.02). Further, a difference in circulating RANTES levels among controls and STEMI subjects, and a relation of serum levels with acute heart failure was observed (pxa0=xa00.03, pxa0=xa00.003, respectively). We found a significant difference when comparing cardiogenic shock patients and controls (pxa0