EMAIL THIS PAGE TO A FRIEND
Journal of autoimmunity
MBD2 regulates TH17 differentiation and experimental autoimmune encephalomyelitis by controlling the homeostasis of T-bet/Hlx axis.
PMID 24934598
Abstract
Unlike genetic alterations, epigenetic modifications are reversible and amenable to pharmacological interventions, which make them appealing targets for clinical therapy. However, little is known about epigenetic regulation in experimental autoimmune encephalomyelitis (EAE). Here we demonstrated that methyl-CpG-binding domain protein 2 (MBD2), an epigenetic regulator, controls autoimmunity and EAE through T-bet/Hlx. Tbx21 and Hlx underwent a DNA methylation turnover upon polarizations and a unique methylation pattern was essential for TH17 development. Loss of Mbd2 resulted in a defect for reading the information encoded by this methylation turnover, which disrupted the homeostasis of T-bet/Hlx axis and suppressed TH17 differentiation. DNA demethylation induced similar effect on helper T cell differentiation. Therefore, Mbd2(-/-) mice were completely protected from EAE. Pathogenic splenocytes isolated from wild-type mice challenged with MOG35-55 could adoptively transfer disease to Mbd2(-/-) mice. In addition, Mbd2(-/-) mice reconstituted with unstimulated wild-type splenocytes developed EAE as wild-type mice did. These data would provide novel insights into epigenetic regulation of EAE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501381
Anti-Cd28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501674
Anti-CD28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405583
Anti-CD28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C7831 Anti-CD28 antibody, Mouse monoclonal, clone CD28.2, purified from hybridoma cell culture
PRS4877
Anti-IL-17 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4887
Anti-IL-17 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2502114
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502115
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA052258
Anti-IL17A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
EHU150651 MISSION® esiRNA, esiRNA human HLX (esiRNA1)
EMU030561 MISSION® esiRNA, esiRNA targeting mouse Hlx (esiRNA1)
SAB4700135 Monoclonal Anti-CD28 antibody produced in mouse, low endotoxin, clone CD28.2, purified immunoglobulin, buffered aqueous solution
SAB4700739 Monoclonal Anti-CD28 antibody produced in mouse, clone JJ319, purified immunoglobulin, buffered aqueous solution
SAB4700751
Monoclonal Anti-CD28 antibody produced in mouse, clone CD28.2, purified immunoglobulin, buffered aqueous solution
SAB3701439
Monoclonal Anti-IL-17A antibody produced in mouse, clone 4H1524.1, purified immunoglobulin
SAB3701458
Monoclonal Anti-IL-17A antibody produced in rat, clone 20B4.G10.F5, purified immunoglobulin