EMAIL THIS PAGE TO A FRIEND
Medical mycology
Uncommon opportunistic yeast bloodstream infections from Qatar.
PMID 24934803
Abstract
Eleven uncommon yeast species that are associated with high mortality rates irrespective of antifungal therapy were isolated from 17/187 (201 episodes) pediatric and elderly patients with fungemia from Qatar. The samples were taken over a 6-year period (January 2004-December 2010). Isolated species included Kluyveromyces marxianus, Lodderomyces elongisporus, Lindnera fabianii, Candida dubliniensis, Meyerozyma guilliermondii, Candida intermedia, Pichia kudriavzevii, Yarrowia lipolytica, Clavispora lusitaniae, Candida pararugosa, and Wickerhamomyces anomalus. Matrix-assisted laser desorption/ionization time-of-flight mass spectrometry provided correct identifications compared with molecular analysis testing of the same isolates. Low minimal inhibitory concentrations were found when isavuconazole and voriconazole were used for all uncommon yeast species evaluated in this study. Resistance to antifungal drugs was low and remained restricted to a few species.