EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
Selective activity of the histone deacetylase inhibitor AR-42 against leukemia stem cells: a novel potential strategy in acute myelogenous leukemia.
PMID 24934933
Abstract
Most patients with acute myelogenous leukemia (AML) relapse and die of their disease. Increasing evidence indicates that AML relapse is driven by the inability to eradicate leukemia stem cells (LSC). Thus, it is imperative to identify novel therapies that can ablate LSCs. Using an in silico gene expression-based screen for compounds evoking transcriptional effects similar to the previously described anti-LSC agent parthenolide, we identified AR-42 (OSU-HDAC42), a novel histone deacetylase inhibitor that is structurally similar to phenylbutyrate, but with improved activity at submicromolar concentrations. Here, we report that AR-42 induces NF-κB inhibition, disrupts the ability of Hsp90 to stabilize its oncogenic clients, and causes potent and specific cell death of LSCs but not normal hematopoietic stem and progenitor cells. Unlike parthenolide, the caspase-dependent apoptosis caused by AR-42 occurs without activation of Nrf-2-driven cytoprotective pathways. As AR-42 is already being tested in early clinical trials, we expect that our results can be extended to the clinic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0667
Parthenolide, ≥98% (HPLC)
C15H20O3