EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Leucocyte expression of complement C5a receptors exacerbates infarct size after myocardial reperfusion injury.
PMID 24935433
Abstract
Early reperfusion is mandatory for the treatment of acute myocardial infarction. This process, however, also induces additional loss of viable myocardium, called ischaemia-reperfusion (IR) injury. Complement activation plays an important role in IR injury, partly through binding of C5a to its major receptor (C5aR). We investigated the role of C5aR on infarct size and cardiac function in a model for myocardial IR injury. BALB/c (WT) mice and C5aR(-/-) mice underwent coronary occlusion for 30 min, followed by reperfusion. Infarct size, determined 24 h after IR, was reduced in C5aR(-/-) mice compared with WT mice (28.5 ± 2.1 vs. 35.7 ± 2.5%, P = 0.017). Bone marrow (BM) chimaera experiments showed that this effect was due to the absence of C5aR on circulating leucocytes, since a similar reduction in infarct size was observed in WT mice with C5aR-deficient BM cells (25.3 ± 2.2 vs. 34.6 ± 2.8%, P < 0.05), but not in C5aR(-/-) mice with WT BM cells. Reduced infarct size was associated with fewer neutrophils, T cells, and macrophages in the infarcted area 24 h after IR in C5aR(-/-) mice, and also with lower levels of Caspase-3/7 indicating less inflammation and apoptosis. Echocardiography 4 weeks after IR showed an improved ejection fraction in C5aR(-/-) mice (25.8 ± 5.5 vs. 19.2 ± 5.4%, P < 0.001). The absence of C5aR on circulating leucocytes reduces infarct size, is associated with reduced leucocyte infiltration and with less apoptosis in the infarcted myocardium, and improves cardiac function in a mouse model of myocardial IR injury. Selective blocking of C5aR might be a promising strategy to prevent myocardial IR injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8877
2,3,5-Triphenyltetrazolium chloride, ≥95%
C19H15ClN4
HPA003563
Anti-C3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA020432
Anti-C3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014520
Anti-C5AR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401040
Anti-C5AR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1405535
Anti-C5R1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
GW20073F Anti-Complement C3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
WH0000718M1
Monoclonal Anti-C3 antibody produced in mouse, clone 5F9, purified immunoglobulin, buffered aqueous solution
SAB1403622
Monoclonal Anti-C3 antibody produced in mouse, clone 5F9, ascites fluid
SAB1402133
Monoclonal Anti-C5R1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution