EMAIL THIS PAGE TO A FRIEND
Asian Pacific journal of cancer prevention : APJCP
Diagnostic and prognostic value of miR-205 in colorectal cancer.
PMID 24935592
Abstract
Emerging evidence has shown associations of microRNA-205 (miR-205) with crucial cell processes such as the epithelial-mesenchymal transition (EMT) and aberrant expression with tumorigenesis in many types of human malignancy. This prospective study characterized the contribution of miR-205 to the colorectal cancer (CRC) tumorigenesis. The real-time reverse transcription-polymerase chain reaction was used to examine miR-205 levels prospectively in 36 pairs of samples of CRC tissue and adjacent noncancerous tissue (>2 cm from cancer tissue). In addition, the relationship between miR-205 levels and clinicopathological features was explored. The capability of miR-205 to function as a tumor marker was also examined. miR-205 expression levels did not show significant changes overall. However, miR-205 was significantly downregulated in a group of CRC samples compared with matched noncancerous tissue samples. Moreover, decreased miR-205 correlated significantly with lymphatic metastasis. A receiver operating characteristic (ROC) curve also showed an optimum cut off point of 1.4?10-3 to distinguish lymphatic metastatic CRCs from non-metastatic CRCs. Interestingly we found lymphatic metastasis in almost 80% of the depressed samples. This study suggested that miR-205 could be reduced in the majority of metastatic CRCs and the risk of CRC metastasis may be predicted by monitoring miR-205 in patient samples collected at the time of the initial diagnosis. Therefore, targeting miR-205 and its potential environmental activators might be a promising therapeutic option to prevent malignant progression toward metastasis.