EMAIL THIS PAGE TO A FRIEND
Transplantation
Late antibody-mediated rejection in renal allografts: outcome after conventional and novel therapies.
PMID 24937198
Abstract
Although several strategies for treating early antibody-mediated rejection (AMR) in kidney transplants have been investigated, evidence on treatment of late AMR manifesting after 6 months is sparse. In this single-center series, we present data on 23 consecutive patients treated for late AMR. Late AMR was diagnosed using Banff 2007 criteria along with presence of donor-specific antibodies (DSA) and acute rise in serum creatinine (SCr). Response to therapy was assessed by improvement in SCr, histologic improvement, and decline in DSA strength. Overall, 17% (4/23) had documented nonadherence while 69% (16/23) had physician-recommended reduction in immunosuppression before AMR. Eighteen patients (78%) were treated with plasmapheresis or low-dose IVIg+rituximab; 11 (49%) with refractory AMR also received one to three cycles of bortezomib. While there was an improvement (P=0.02) in mean SCr (2.4 mg/dL) at the end of therapy compared with SCr at the time of diagnosis (2.9 mg/dL), this improvement was not sustained at most recent follow-up. Eleven (48%) patients had no histologic resolution on follow-up biopsy. Lack of histologic response was associated with older patients (odds ratio [OR]=3.17; P=0.04), presence of cytotoxic DSA at time of diagnosis (OR=200; P=0.04), and severe chronic vasculopathy (cv≥2) on index biopsy (OR=50; P=0.06). A major setting in which late AMR occurred in our cohort was reduction or change in immunosuppression. Our data demonstrate an inadequate response of late AMR to current and novel (bortezomib) therapies. The benefits of therapy need to be counterweighed with potential adverse effects especially in older patients, large antibody loads, and chronic allograft vasculopathy.