EMAIL THIS PAGE TO A FRIEND
Oncotarget
Chemopreventive and chemotherapeutic effect of dietary supplementation of vitamin D on cholangiocarcinoma in a Chemical-Induced animal model.
PMID 24939880
Abstract
Intrahepatic cholangiocarcinoma (ICC) is an aggressive cancer. Vitamin D, a pro-hormone, is getting popular due to its hormone-like functions after converted to its active form, 1α,25(OH)2D3. Here, we show that dietary supplementation with 6 IU/g of vitamin D greatly suppressed ICC initiation and progression without apparent toxicity in a chemically induced rat model. Microarray analysis of rat ICC tissues showed vitamin D supplementation modulated the expressions of several unique genes, including lipocalin 2 (Lcn2), confirmed by RT-qPCR and immunohistochemical (IHC) staining. Further, 53 of 80 human ICC specimens (66%) exhibited high LCN2 expression and LCN2 knockdown in SNU308 cells decreased cell growth and migration, suggesting LCN2 be an oncogene in human ICC. As human ICC SNU1079 cells were treated by 1α,25(OH)2D3, LCN2 expression and cell proliferation were attenuated. The downregulation of LCN2 expression was blunted when vitamin D receptor (VDR) was knocked down, implicating that the in vivo Lcn2 downregulation is a direct consequence of vitamin D supplementation Our results support the prevailing concept that vitamin D status is negatively associated with cancer incidence and mortality and suggest LCN2 may be a potential target against ICC. Further studies of application of vitamin D or its analog against ICC are warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700855 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700857 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700851 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700847 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700854 Anti-Rabbit IgG (Fc specific)-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700856 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700850 Anti-Rabbit IgG (Fc specific)-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700846 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600006 Anti-Rabbit IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB3700950 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700951 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700947 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700945 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700973 Anti-Rabbit IgG (H+L), F(ab) fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700971 Anti-Rabbit IgG (H+L), F(ab) fragment-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700969 Anti-Rabbit IgG (H+L), F(ab) fragment-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600007 Anti-Rabbit IgG (H+L), highly cross-adsorbed, Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
A3937 Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A8025 Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A9919 Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A7539 Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A0418 Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum
G7402
Anti-Rabbit IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
G7277 Anti-Rabbit IgG (whole molecule)−Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 5 nm (colloidal gold)
G3779 Anti-Rabbit IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
T5268 Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
18772 Anti-Rabbit IgG - Atto 488 antibody produced in goat, whole molecule, 1 mg/mL protein
77671 Anti-Rabbit IgG - Atto 594 antibody produced in goat
41176 Anti-Rabbit IgG - Atto 633 antibody produced in goat, 1 mg/mL IgG
78519 Anti-Rabbit IgG - Atto 655 antibody produced in goat, ~1 mg/mL IgG