EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Thimerosal induces apoptotic and fibrotic changes to kidney epithelial cells in vitro.
PMID 24942245
Abstract
Thimerosal is an ethyl mercury-containing compound used mainly in vaccines as a bactericide. Although the kidney is a key target for mercury toxicity, thimerosal nephrotoxicity has not received the same attention as other mercury species. The aim of this study was to determine the potential cytotoxic mechanisms of thimerosal on human kidney cells. Human kidney proximal tubular epithelial (HK2) cells were exposed for 24 h to thimerosal (0-2 µM), and assessed for cell viability, apoptosis, and cell proliferation; expression of proteins Bax, nuclear factor-κB subunits, and transforming growth factor-β1 (TGFβ1); mitochondrial health (JC-1, MitoTracker Red CMXRos); and fibronectin levels (enzyme-linked immunosorbent assay). Thimerosal diminished HK2 cell viability and mitosis, promoted apoptosis, impaired the mitochondrial permeability transition, enhanced Bax and TGFβ1 expression, and augmented fibronectin secretion. This is the first report about kidney cell death and pro-fibrotic mechanisms promoted by thimerosal. Collectively, these in vitro results demonstrate that (1) thimerosal induces kidney epithelial cell apoptosis via upregulating Bax and the mitochondrial apoptotic pathway, and (2) thimerosal is a potential pro-fibrotic agent in human kidney cells. We suggest that new evidence on toxicity as well as continuous surveillance in terms of fibrogenesis is required concerning thimerosal use.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MHS32
Hematoxylin Solution, Mayer’s