EMAIL THIS PAGE TO A FRIEND
European journal of clinical investigation
FGF-23 dysregulates calcium homeostasis and electrophysiological properties in HL-1 atrial cells.
PMID 24942561
Abstract
Fibroblast growth factor (FGF)-23 is a key regulator of phosphate homeostasis. Higher FGF-23 levels are correlated with poor outcomes in cardiovascular diseases. FGF-23 can produce cardiac hypertrophy and increase intracellular calcium, which can change cardiac electrical activity. However, it is not clear whether FGF-23 possesses arrhythmogenic potential through calcium dysregulation. Therefore, the purposes of this study were to evaluate the electrophysiological effects of FGF-23 and identify the underlying mechanisms. Patch clamp, confocal microscope with Fluo-4 fluorescence, and Western blot analyses were used to evaluate the electrophysiological characteristics, calcium homeostasis and calcium regulatory proteins in HL-1 atrial myocytes with and without FGF-23 (10 and 25xa0ng/mL) incubation for 24xa0h. FGF-23 (25xa0ng/mL) increased L-type calcium currents, calcium transient and sarcoplasmic reticulum Ca(2+) contents in HL-1 cells. FGF-23 (25xa0ng/mL)-treated cells (nxa0=xa014) had greater incidences (57%, 17% and 15%, Pxa0