EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
Melanocortin 4 receptor activation inhibits presynaptic N-type calcium channels in amygdaloid complex neurons.
PMID 24943127
Abstract
The melanocortin 4 receptor (MC4R) is a G protein-coupled receptor involved in food intake and energy expenditure regulation. MC4R activation modifies neuronal activity but the molecular mechanisms by which this regulation occurs remain unclear. Here, we tested the hypothesis that MC4R activation regulates the activity of voltage-gated calcium channels and, as a consequence, synaptic activity. We also tested whether the proposed effect occurs in the amygdala, a brain area known to mediate the anorexigenic actions of MC4R signaling. Using the patch-clamp technique, we found that the activation of MC4R with its agonist melanotan II specifically inhibited 34.5 ± 1.5% of N-type calcium currents in transiently transfected HEK293 cells. This inhibition was concentration-dependent, voltage-independent and occluded by the Gαs pathway inhibitor cholera toxin. Moreover, we found that melanotan II specifically inhibited 25.9 ± 2.0% of native N-type calcium currents and 55.4 ± 14.4% of evoked inhibitory postsynaptic currents in mouse cultured amygdala neurons. In vivo, we found that the MC4R agonist RO27-3225 increased the marker of cellular activity c-Fos in several components of the amygdala, whereas the N-type channel blocker ω conotoxin GVIA increased c-Fos expression exclusively in the central subdivision of the amygdala. Thus, MC4R specifically inhibited the presynaptic N-type channel subtype, and this inhibition may be important for the effects of melanocortin in the central subdivision of the amygdala.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21144 Anti-c-Fos antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
F7799
Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
F137 Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
HPA018531
Anti-C-FOS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100832
Anti-FOS (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100833
Anti-FOS (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2502116
Anti-FOS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100639
Anti-FOS antibody produced in rabbit, IgG fraction of antiserum
SAB2104185
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500995
Anti-FOS antibody produced in rabbit, affinity isolated antibody
C7527
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
C5H14ClNO
C7017
Choline chloride, ≥99%
C5H14ClNO
1133547
Choline chloride, United States Pharmacopeia (USP) Reference Standard
C5H14ClNO
C1879
Choline chloride, ≥98%
C5H14ClNO
26978
Choline chloride, BioUltra, ≥99.0% (AT)
C5H14ClNO
PHR1251
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
C5H14ClNO
SAB5300463
Monoclonal Anti-FOS antibody produced in mouse, clone 2G2, ascites fluid
SAB1402201
Monoclonal Anti-FOS antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution