EMAIL THIS PAGE TO A FRIEND
Mayo Clinic proceedings
Imaging Pandora's Box: incidental findings in elderly patients evaluated for transcatheter aortic valve replacement.
PMID 24943693
Abstract
To determine the frequency and clinical impact of noncardiovascular incidental findings (IFs) detected on preoperative computed tomographic angiography (CTA) of the chest/abdomen/pelvis performed in elderly patients with severe aortic stenosis being considered for transcatheter aortic valve replacement (TAVR). The CTA studies for 424 consecutive patients being evaluated for TAVR between January 1, 2009, through January 24, 2012, were reviewed for noncardiovascular IFs (62.0% male; median ± SD age, 82 ± 8.3 years). The electronic medical record was reviewed to assess for subsequent clinical management and survival. Potentially pathologic IFs (PPIFs) were present in 285 patients (67.2%). The mean ± SD number of PPIFs per patient was 1.1 ± 1.0 (range, 0-4). Factors associated with higher numbers of PPIFs were reduced ejection fraction (P=.02) and history of smoking (P=.06). Potentially pathologic incidental findings prompted clinical work-up in 39 patients (9.2%) and delayed or canceled treatment plans for aortic stenosis in 7 patients (1.7%). The number needed to image to diagnose a new malignancy or medical condition was 19. The number of PPIFs was predictive of poor overall survival before (hazard ratio, 1.58; 95% CI, 1.31-1.88) and after (hazard ratio, 1.45; 95% CI, 1.19-1.76) adjustment for baseline clinical variables (P<.001 for both). This investigation found that PPIFs are common in elderly patients undergoing TAVR-CTA scans and, once discovered, commonly generate further clinical evaluation. Higher numbers of PPIFs may be predictive of poorer survival, but further study is required to guide the appropriateness of pursuing diagnostic evaluations for asymptomatic PPIFs in this elderly population.