EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
The SGLT2 inhibitor empagliflozin ameliorates early features of diabetic nephropathy in BTBR ob/ob type 2 diabetic mice with and without hypertension.
PMID 24944269
Abstract
Diabetic nephropathy is the leading cause of end-stage renal disease in humans in the Western world. The recent development of Na+-glucose cotransporter 2 (SGLT2) inhibitors offers a new antidiabetic therapy via enhanced glucose excretion. Whether this strategy exerts beneficial effects on the development of type 2 diabetic nephropathy is still largely unclear. We investigated the effects of the specific SGLT2 inhibitor empagliflozin in BTBR.Cg-Lep/WiscJ (BTBR ob/ob) mice, which spontaneously develop type 2 diabetic nephropathy. In the first experiment, BTBR ob/ob mice received either a diet containing 300 ppm empagliflozin or equicaloric placebo chow for 12 wk. In the second experiment, BTBR ob/ob mice received 1 μg·kg body wt(-1)·day(-1) ANG II to induce arterial hypertension and were separated into the same two diet groups for 6 wk. In both experiments, empagliflozin treatment enhanced glucosuria, thereby lowering blood glucose. Independently of hypertension, empagliflozin reduced albuminuria in diabetic mice. However, empagliflozin treatment affected diabetes-related glomerular hypertrophy, markers of renal inflammation, and mesangial matrix expansion only in BTBR ob/ob mice without hypertension. In summary, empagliflozin demonstrated significant antihyperglycemic effects, differentially ameliorating early features of diabetic nephropathy in BTBR ob/ob mice with and without hypertension.