EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
The cholinergic basal forebrain in the ferret and its inputs to the auditory cortex.
PMID 24945075
Abstract
Cholinergic inputs to the auditory cortex can modulate sensory processing and regulate stimulus-specific plasticity according to the behavioural state of the subject. In order to understand how acetylcholine achieves this, it is essential to elucidate the circuitry by which cholinergic inputs influence the cortex. In this study, we described the distribution of cholinergic neurons in the basal forebrain and their inputs to the auditory cortex of the ferret, a species used increasingly in studies of auditory learning and plasticity. Cholinergic neurons in the basal forebrain, visualized by choline acetyltransferase and p75 neurotrophin receptor immunocytochemistry, were distributed through the medial septum, diagonal band of Broca, and nucleus basalis magnocellularis. Epipial tracer deposits and injections of the immunotoxin ME20.4-SAP (monoclonal antibody specific for the p75 neurotrophin receptor conjugated to saporin) in the auditory cortex showed that cholinergic inputs originate almost exclusively in the ipsilateral nucleus basalis. Moreover, tracer injections in the nucleus basalis revealed a pattern of labelled fibres and terminal fields that resembled acetylcholinesterase fibre staining in the auditory cortex, with the heaviest labelling in layers II/III and in the infragranular layers. Labelled fibres with small en-passant varicosities and simple terminal swellings were observed throughout all auditory cortical regions. The widespread distribution of cholinergic inputs from the nucleus basalis to both primary and higher level areas of the auditory cortex suggests that acetylcholine is likely to be involved in modulating many aspects of auditory processing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0639
6α-Methylprednisolone, ≥98%
C22H30O5
46436
6α-Methylprednisolone, VETRANAL, analytical standard
C22H30O5
AB143
Anti-Choline Acetyltransferase (ChAT) Antibody, Anti-Choline Acetyltransferase (ChAT) Antibody is an antibody against Choline Acetyltransferase (ChAT) for use in IH, IH(P), IP & WB with more than 70 product citations.
A0132
Atropine, ≥99% (TLC), powder
C17H23NO3
37019
Atropine, analytical standard
C17H23NO3
Y0000878
Atropine, European Pharmacopoeia (EP) Reference Standard
C17H23NO3
Y0000845
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H23NO3
C4522
Cimetidine
C10H16N6S
PHR1075
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H16N6S
C2175000
Cimetidine, European Pharmacopoeia (EP) Reference Standard
C10H16N6S
1134062
Cimetidine, United States Pharmacopeia (USP) Reference Standard
C10H16N6S
Y0001130
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
C10H16N6S
Y0001146
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
C10H16N6S
62860
DL-Lysine, purum, ≥95.0% (NT)
C6H14N2O2
Y0000796
Fluorescein, European Pharmacopoeia (EP) Reference Standard
C20H12O5
M2392
Meprobamate, analytical standard
C9H18N2O4
M0400000
Meprobamate, European Pharmacopoeia (EP) Reference Standard
C9H18N2O4
M-039
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C9H18N2O4
M1750000
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard
C22H30O5
1435003
Methylprednisolone, United States Pharmacopeia (USP) Reference Standard
C22H30O5
Y0001602
Methylprednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H30O5
G6539
Monoclonal Anti-Green Fluorescent Protein (GFP) antibody produced in mouse, clone GFP-20, ascites fluid
63183
N-(3-Maleimidopropionyl)biocytin, ≥75% (HPLC)
C23H33N5O7S
95660
o-Xylene, analytical standard
C8H10
PHR1312
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10