PloS one
Phospholipase C-related catalytically inactive protein (PRIP) regulates lipolysis in adipose tissue by modulating the phosphorylation of hormone-sensitive lipase.
PMID 24945349
Abstract
Phosphorylation of hormone-sensitive lipase (HSL) and perilipin by protein kinase A (PKA) promotes the hydrolysis of lipids in adipocytes. Although activation of lipolysis by PKA has been well studied, inactivation via protein phosphatases is poorly understood. Here, we investigated whether phospholipase C-related catalytically inactive protein (PRIP), a binding partner for protein phosphatase 1 and protein phosphatase 2A (PP2A), is involved in lipolysis by regulating phosphatase activity. PRIP knockout (PRIP-KO) mice displayed reduced body-fat mass as compared with wild-type mice fed with standard chow ad libitum. Most other organs appeared normal, suggesting that mutant mice had aberrant fat metabolism in adipocytes. HSL in PRIP-KO adipose tissue was highly phosphorylated compared to that in wild-type mice. Starvation of wild-type mice or stimulation of adipose tissue explants with the catabolic hormone, adrenaline, translocated both PRIP and PP2A from the cytosol to lipid droplets, but the translocation of PP2A was significantly reduced in PRIP-KO adipocytes. Consistently, the phosphatase activity associated with lipid droplet fraction in PRIP-KO adipocytes was significantly reduced and was independent of adrenaline stimulation. Lipolysis activity, as assessed by measurement of non-esterified fatty acids and glycerol, was higher in PRIP-KO adipocytes. When wild-type adipocytes were treated with a phosphatase inhibitor, they showed a high lipolysis activity at the similar level to PRIP-KO adipocytes. Collectively, these results suggest that PRIP promotes the translocation of phosphatases to lipid droplets to trigger the dephosphorylation of HSL and perilipin A, thus reducing PKA-mediated lipolysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000882
Adrenaline, European Pharmacopoeia (EP) Reference Standard
C9H13NO3
SAB1406995
Anti-CLEC10A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021937
Anti-CLEC10A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB4501762
Anti-HSL antibody produced in rabbit, affinity isolated antibody
SAB4501763
Anti-HSL antibody produced in rabbit, affinity isolated antibody
PRS3965
Anti-Lipe antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900275
Anti-LIPE antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW22067 Anti-MGLL antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407139
Anti-MGLL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011993
Anti-MGLL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA011994
Anti-MGLL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408669
Anti-MGLL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500641
Anti-Monoglyceride lipase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA027863
Anti-NPY4R antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
P1998 Anti-Perilipin A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503879
Anti-phospho-HSL (pSer552) antibody produced in rabbit, affinity isolated antibody
SAB4503821
Anti-phospho-HSL (pSer855/554) antibody produced in rabbit, affinity isolated antibody
AV42057
Anti-PLIN antibody produced in rabbit, IgG fraction of antiserum
HPA024299
Anti-PLIN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407769
Anti-PNPLA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411126
Anti-PNPLA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22072A Anti-PPA1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22072B Anti-PPA1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV48396
Anti-PPA1 antibody produced in rabbit, IgG fraction of antiserum
HPA020096
Anti-PPA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA019878
Anti-PPA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
GW22549 Anti-PPP1CC antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406283
Anti-PPP1CC antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701938
Anti-ppp1cc antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1300598
Anti-PPP2R1A (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution