PloS one
Circulating autoantibodies to endothelial progenitor cells: binding characteristics and association with risk factors for atherosclerosis.
PMID 24945945
Abstract
Endothelial progenitor cells (EPC) are committed to transform into EC promoting vasculogenic ischemic repair. Anti-endothelial cells (AECA) have been described in various disorders with an associated vascular damage. Herein, we explored a novel circulating population of IgG reactive with EPC, in patients with differential risk profile for atherosclerotic vascular disease. A novel cyto-ELISA system was established where the coated cells were late outgrowth EPC. Levels of anti-EPC antibodies were determined in 100 subjects and differential risk score for atherosclerosis, as well as to circulating EPC levels and the inflammatory markers IL-6 and C-reactive protein. To study endothelial cell (EC) activating properties, sera were tested for their ability to induce VCAM-1 expression in a cell ELISA system. Detectable levels of anti-EPC antibodies, that correlated with age, Framingham risk score and CRP concentrations but did not associate with levels of LDL, HDL, hypertension or diabetes, were detected. Anti-EPC antibodies were distinct from EC binding antibodies as shown by competitive inhibition studies, and have been positively correlated with the extent of EC activation manifested by in vitro VCAM-1 expression. This is the first study showing a newly defined subgroup of self-antibodies binding EPC and associating positively with the Framingham risk score. Further studies are required to characterize and test this interesting subset of EPC binding autoantibodies and their potential significance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8542 Anti-Human IgG (Fab specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701267 Anti-Human IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701277 Anti-Human IgG (Fc specific), highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9544 Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3312
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A5403 Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3187 Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A3150 Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A3188 Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A1543 Anti-Human IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB3701239 Anti-Human IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701248 Anti-Human IgG F(ab′)2-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
RAB0507
Human VEGF ELISA Kit, for serum, plasma, cell culture supernatants and urine
RAB0508
Human VEGF ELISA Kit, for cell and tissue lysates
RAB0328
Human VEGF R2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0195
Human VEGF R3 ELISA Kit, for serum, plasma, cell culture supernatant, urine
RAB0509
Mouse VEGF ELISA Kit, for serum, plasma, cell culture supernatant
RAB0510
Mouse VEGF ELISA Kit, for cell and tissue lysates
RAB0192
Mouse VEGF R1 ELISA Kit, for serum, plasma and cell culture supernatants
RAB0511
Rat VEGF ELISA Kit, for serum, plasma and cell culture supernatant
RAB0512
Rat VEGF ELISA Kit, for cell and tissue lysates