EMAIL THIS PAGE TO A FRIEND
Journal of translational medicine
Adoptive immunotherapy with MUC1-mRNA transfected dendritic cells and cytotoxic lymphocytes plus gemcitabine for unresectable pancreatic cancer.
PMID 24947606
Abstract
We previously reported the clinical efficacy of adoptive immunotherapy (AIT) with dendritic cells (DCs) pulsed with mucin 1 (MUC1) peptide and cytotoxic T lymphocytes (CTLs). We also reported that gemcitabine (GEM) enhances anti-tumor immunity by suppressing regulatory T cells. Therefore, in the present study, we performed combination therapy with AIT and GEM for patients with unresectable or recurrent pancreatic cancer. Forty-two patients with unresectable or recurrent pancreatic cancer were treated. DCs were generated by culture with granulocyte macrophage colony-stimulating factor and interleukin-4 and then exposed to tumor necrosis factor-α. Mature DCs were transfected with MUC1-mRNA by electroporation (MUC1-DCs). MUC1-CTLs were induced by co-culture with YPK-1, a human pancreatic cancer cell line, and then with interleukin-2. Patients were treated with GEM, while MUC1-DCs were intradermally injected, and MUC1-CTLs were intravenously administered. Median survival time (MST) was 13.9 months, and the 1-year survival rate was 51.1%. Of 42 patients, one patient had complete response (2.4%), three patients had partial response (7.1%) and 22 patients had stable disease (52.4%). The disease control ratio was 61.9%. The MST and 1-year survival rate of 35 patients who received more than 1 × 10(7) MUC1-DCs per injection was 16.1 months and 60.3%, respectively. Liver metastasis occurred in only 5 patients among 35 patients without liver metastasis before treatment. There were no severe toxicities associated with AIT. AIT with MUC1-DCs and MUC1-CTLs plus GEM may be a feasible and effective treatment for pancreatic cancer.