EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Histamine monolith versatility to purify supercoiled plasmid deoxyribonucleic acid from Escherichia coli lysate.
PMID 24947891
Abstract
Preparation of high quantities of supercoiled plasmid DNA of pharmaceutical grade purity is a research area where intensive investigation is being performed. From this standpoint, several downstream methods have been proposed, among them the monolithic chromatographic strategies owing to excellent mass transfer properties of monolithic supports and their high binding capacity for large biomolecules. The present study explores the physicochemical properties of histamine ligand in a supercoiled plasmid DNA purification process from an Escherichia coli clarified lysate, where the emphasis is given to the elution strategy that allows higher selectivity and efficient removal of other impurities besides the open circular isoform. The combination of high NaCl concentration and acidic pH allowed the elimination of 89% of RNA during the preparative loading of the lysate sample. The results of the purification strategy with ascending sodium chloride gradient revealed that 97% of supercoiled plasmid DNA was recovered with a purity degree of 99%. In addition, using a combined purification strategy with ascending sodium chloride (capture step) and then descending ammonium sulfate (polishing step) gradient, it was achieved a lower supercoiled plasmid DNA recovery yield of 79% with a purity degree of 92%, although the dynamic binding capacity under these conditions was higher than in the previous strategy. A significant reduction of host contents, such as proteins, RNA and genomic DNA, was obtained in both purification strategies. Accordingly, histamine is a useful and versatile ligand that allows the desirable supercoiled plasmid purification with high yield and purity level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RES1427A-A7
Ammonium sulfate, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H8N2O4S
A4418
Ammonium sulfate, for molecular biology, ≥99.0%
H8N2O4S
A6387
Ammonium sulfate, ReagentPlus®, ≥99.0%
H8N2O4S
A3920
Ammonium sulfate, plant cell culture tested, ≥99.0%
H8N2O4S
09978
Ammonium sulfate, BioUltra, ≥99.5% (T)
H8N2O4S
A2939
Ammonium sulfate, BioXtra, ≥99.0%
H8N2O4S
204501
Ammonium sulfate, 99.999% trace metals basis
H8N2O4S
59845
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
H8N2O4S
451975
Ammonium sulfate-14N2 solution, 99.99 atom % 14N, 40 wt. % in H2O
H814N2O4S
485411
Ammonium-14N2 sulfate, 99.99 atom % 14N
H814N2O4S
485306
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
H814N2O4S
608602
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
H814N216O4S
38730 Citrate standard for IC, 1000 mg/L, analytical standard
C6H8O7
94676
Citric acid, certified reference material, TraceCERT®
C6H8O7
46933
Citric acid, analytical standard
C6H8O7
1134368
Citric acid, United States Pharmacopeia (USP) Reference Standard
C6H8O7
A1202000
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
C6H8O7
PHR1071
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
C6H8O7
59964
Histamine, analytical standard
C5H9N3
H7125
Histamine, ≥97.0%
C5H9N3
53290
Histamine, base, ≥97.0% (NT)
C5H9N3
T1378
Sigma 7-9®, ≥99% (titration), crystalline
C4H11NO3
RDD008
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
C4H11NO3
T6066
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration)
C4H11NO3
T6791
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
C4H11NO3
T4661
Trizma® base, ≥99.9% (titration), crystalline
C4H11NO3
93350
Trizma® base, puriss. p.a., ≥99.7% (T)
C4H11NO3
33742
Trizma® base, puriss. p.a., buffer substance, ≥99.5%
C4H11NO3
04577
Trizma® base, tested according to Ph.Eur.
C4H11NO3
93440
Trizma® base, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
C4H11NO3