EMAIL THIS PAGE TO A FRIEND
Cell biochemistry and biophysics
Edaravone protects neurons in the rat substantia nigra against 6-hydroxydopamine-induced oxidative stress damage.
PMID 24948472
Abstract
To investigate the mechanism of the neuroprotective effect of edaravone in substantia nigra (SN) of the 6-OHDA-induced rat model of Parkinson's disease. Animal model of Parkinson's disease was induced in male Sprague-Dawley rats by injecting 6-OHDA into the left medial forebrain bundle. Subsequently, rats were intraperitoneally injected with 0.3, 1, or 3 mg/kg of edaravone for 14 days or with 3 mg/kg edaravone for 14 days followed by 14 days of no treatment. We evaluated the effect of edaravone on the rotational and normal behavior of the rats, and on the number of tyrosine hydroxylase (TH)-positive cells, the amount of Nissl bodies, and the levels of glutathione (GSH), and malondialdehyde (MDA) in the SN. Edaravone treatment at 3 mg/kg significantly reduced apomorphine-induced rotational behavior (P < 0.01), improved the spontaneous behavior, prevented the decrease in the levels of TH-positive cells, Nissl bodies and GSH, and inhibited the increase in the levels of MDA (P < 0.05) in SN of rats with 6-OHDA-induced PD. Edaravone exerted a long-term neuroprotective effects in 6-OHDA-induced PD animal model by attenuating changes in the levels of GSH and MDA in SN, caused by oxidative stress. Edaravone prevented 6-OHDA-induced behavioral changes and de-pigmentation of SN that results from the loss of dopaminergic neurons.