EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Evidence for the involvement of spinal cord-inhibitory and cytokines-modulatory mechanisms in the anti-hyperalgesic effect of hecogenin acetate, a steroidal sapogenin-acetylated, in mice.
PMID 24950436
Abstract
Hecogenin is a steroidal sapogenin largely drawn from the plants of the genus Agave, commonly known as 'sisal', and is one of the important precursors used by the pharmaceutical industry for the synthesis of steroid hormones. Hecogenin acetate (HA) is a steroidal sapogenin-acetylated that produces antinociceptive activity. Thus, we evaluate the antihyperalgesic profile of HA in mice in inflammatory models, as well as its possible involvement with c-fos expression on spinal cord area and cytokines to produces analgesic profile. Acute pretreatment with HA (5, 10, or 20 mg/kg; i.p.) inhibited the development of mechanical hyperalgesia induced by carrageenan, TNF-α, dopamine and PGE2. Additionally, the immunofluorescence data demonstrated that acute pretreatment with HA, at all doses tested, significantly inhibited Fos-like expression in the spinal cord dorsal horn normally observed after carrageenan-inflammation. Moreover, HA did not affect the motor performance of the mice as tested in the Rota rod test. This antinociceptive profile seems to be related, at least in part, to a reduction of pro-inflammatory cytokines, as IL-1β. The present results suggest that HA attenuates mechanical hyperalgesia by blocking the neural transmission of pain at the spinal cord levels and by cytokines-inhibitory mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4388
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
C2HBrClF3
SAB3700864 Anti-Rabbit IgG (Fc specific)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700865 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700862 Anti-Rabbit IgG (Fc specific)-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700860 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700979 Anti-Rabbit IgG (H+L), F(ab) fragment-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700977 Anti-Rabbit IgG (H+L), F(ab) fragment-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700975 Anti-Rabbit IgG (H+L), F(ab) fragment-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB4600003 Anti-Rabbit IgG (H+L), highly cross-adsorbed, Biotin antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB3700935 Anti-Rabbit IgG (H+L)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700936 Anti-Rabbit IgG (H+L)-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700933 Anti-Rabbit IgG (H+L)-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700925 Anti-Rabbit IgG (H+L)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
H4274
Hecogenin acetate, ≥90%
C29H44O5