EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
Linking T-cell receptor sequence to functional phenotype at the single-cell level.
PMID 24952902
Abstract
Although each T lymphocyte expresses a T-cell receptor (TCR) that recognizes cognate antigen and controls T-cell activation, different T cells bearing the same TCR can be functionally distinct. Each TCR is a heterodimer, and both α- and β-chains contribute to determining TCR antigen specificity. Here we present a methodology enabling integration of information about TCR specificity with information about T cell function. This method involves sequencing of TCRα and TCRβ genes, and amplifying functional genes characteristic of different T cell subsets, in single T cells. Because this approach retains information about individual TCRα-TCRβ pairs, TCRs of interest can be expressed and used in functional studies, for antigen discovery, or in therapeutic applications. We apply this approach to study the clonal ancestry and differentiation of T lymphocytes infiltrating a human colorectal carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405577
Anti-CD8A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037756
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407427
Anti-FOXP3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410887
Anti-FOXP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4700360 Monoclonal Anti-CD25 antibody produced in mouse, clone MEM-181, purified immunoglobulin, buffered aqueous solution
SAB4700365 Monoclonal Anti-CD25 antibody produced in mouse, clone MEM-140, purified immunoglobulin, buffered aqueous solution
SAB1305503
MONOCLONAL ANTI-CD25 antibody produced in mouse, clone 7G7B6, crude ascites, buffered aqueous solution
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
C1805 Monoclonal Anti-CD4 antibody produced in mouse, clone Q4120, purified immunoglobulin, buffered aqueous solution
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
C7423
Monoclonal Anti-CD8 antibody produced in mouse, clone UCHT-4, purified immunoglobulin, buffered aqueous solution
SAB4700084 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-31, purified immunoglobulin, buffered aqueous solution
SAB4700089 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-87, purified immunoglobulin, buffered aqueous solution
SAB1402142
Monoclonal Anti-CD8A antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution