EMAIL THIS PAGE TO A FRIEND
Steroids
Novel approach to the preparation of hemisuccinates of steroids bearing tertiary alcohol group.
PMID 24954059
Abstract
17β-O-Hemisuccinates of typical representatives of Anabolic-Androgenic Steroids, 17β-hydroxy-17-methylandrostan-4-en-3-one, 17β-hydroxy-17-methyl-2-oxa-5α-androstan-3-one, 17β-hydroxy-17-methyl-5α-androstano-[3,2-c]pyrazole, were prepared. Several methods for the hemisuccinate preparation were tested. The indirect method using 1-ethyl-3-(dimethylaminopropyl)carbodiimide coupling reagent to form an ester bond of steroid with 2-(trimethylsilyl)ethyl hydrogen butanedioate was finally applied. Using the selectively removable protecting group, the desired hemisuccinates of steroids bearing tertiary alcohol group were obtained.