PloS one
Mangiferin attenuates TH1/TH2 cytokine imbalance in an ovalbumin-induced asthmatic mouse model.
PMID 24955743
Abstract
Mangiferin is a major bioactive ingredient in Mangifera indica Linn. (Anacardiaceae) leaves. Aqueous extract of such leaves have been used as an indigenous remedy for respiratory diseases like asthma and coughing in traditional Chinese medicine. However, underlying molecular mechanisms of mangiferin on anti-asthma remain unclear. In our present study, we investigated the anti-asthmatic effect of mangiferin on Th1/Th2 cytokine profiles and explored its underlying immunoregulatory mechanism in mouse model of allergic asthma. Mangiferin significantly reduced the total inflammatory cell counts and eosinophil infiltration, decreased the production of ovalbumin-specific IgE in serum and PGD2 in BALF. The antibody array analysis showed that mangiferin down-regulated the levels of one group of cytokines/chemokines including Th2-related IL-4, IL-5, IL-13, and others IL-3, IL-9, IL-17, RANTES, TNF-α, but simultaneously up-regulated Th1-related IFN-γ, IL-2 and IL-10 and IL-12 expression in serum. Thus it attenuates the imbalance of Th1/Th2 cells ratio by diminishing the abnormal mRNA levels of Th1 cytokines (IFN-γ and IL-12) and Th2 cytokines (IL-4, IL-5 and IL-13). Finally, mangiferin substantially inhibited the activation and expression of STAT-6 and GATA-3 in excised lung tissues. Our results suggest that mangiferin can exert anti-asthmatic effect. The underlying mechanism may attribute to the modulation of Th1/Th2 cytokine imbalance via inhibiting the STAT6 signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300328
Anti-STAT4 (Ab-693) antibody produced in rabbit, affinity isolated antibody
GW22786 Anti-STAT4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501629
Anti-STAT4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501630
Anti-STAT4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001860
Anti-STAT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33376
Anti-STAT4 antibody produced in rabbit, IgG fraction of antiserum
S6433 Anti-STAT6 (787-804) antibody produced in rabbit, whole antiserum
SAB4300331
Anti-STAT6 (Ab-641) antibody produced in rabbit, affinity isolated antibody
SAB4300332
Anti-STAT6 (Ab-645) antibody produced in rabbit, affinity isolated antibody
AV31048
Anti-STAT6 (AB1) antibody produced in rabbit, affinity isolated antibody
AV38257
Anti-STAT6 (AB2) antibody produced in rabbit, affinity isolated antibody
GW22760 Anti-STAT6 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406489
Anti-STAT6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001861
Anti-STAT6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502882
Anti-STAT6 antibody produced in rabbit, affinity isolated antibody
WH0006775M1
Monoclonal Anti-STAT4 antibody produced in mouse, clone 1C2-1C12, purified immunoglobulin, buffered aqueous solution
WH0006778M1
Monoclonal Anti-STAT6 antibody produced in mouse, clone 6C10, purified immunoglobulin, buffered aqueous solution
SAB1404418
Monoclonal Anti-STAT6 antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB5300363 Monoclonal Anti-STAT6 antibody produced in mouse, clone 7D3, ascites fluid