EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
High cAMP attenuation of insulin-stimulated meiotic G2-M1 transition in zebrafish oocytes: interaction between the cAMP-dependent protein kinase (PKA) and the MAPK3/1 pathways.
PMID 24956082
Abstract
High intra-cellular cyclic nucleotide (cAMP) ensures prophase-I arrest and prevent steroid-induced meiotic G2-M1 transition in full-grown oocytes; however, relatively less information is available for cAMP regulation of growth factor-stimulated signalling events in the oocyte model. Here using zebrafish oocytes, we show that priming with dibutyryl cAMP (dbcAMP) or cAMP modulators, e.g. adenylate cyclase activator, forskolin or phosphodiesterase inhibitors (IBMX/cilostamide) block insulin action on germinal vesicle breakdown (GVBD) and histone H1 kinase activation. Though high cAMP priming attenuates insulin-induced MAPK3/1 (ERK1/2) phosphorylation (activation), following 2h of insulin stimulation it fails to block MAPK activation and GVBD. Further, insulin stimulation promotes down regulation of phospho-PKAc (inactivation) and PKA inhibition by H89/PKI-(6-22)-amide overcomes negative regulation by cAMP and induces GVBD and MAPK activation. Moreover, MEK1/2 inhibitor U0126 has no influence on H89-induced GVBD; however, it delays GVBD response in insulin-stimulated oocytes. MAPK activation by okadaic acid (OA) promotes GVBD; however, high dbcAMP abrogates OA action suggesting cross-talk between cAMP/PKA and MAPK-mediated signalling pathways may contribute significantly in maturing zebrafish oocyte.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4502571
Anti-C-RAF antibody produced in rabbit, affinity isolated antibody
SAB4502573
Anti-C-RAF antibody produced in rabbit, affinity isolated antibody
SAB4502575
Anti-C-RAF antibody produced in rabbit, affinity isolated antibody
SAB4502576
Anti-C-RAF antibody produced in rabbit, affinity isolated antibody
SAB4502577
Anti-C-RAF antibody produced in rabbit, affinity isolated antibody
SAB4502578
Anti-C-RAF antibody produced in rabbit, affinity isolated antibody
SAB4300493
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
SAB4300494
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
GW22781 Anti-MAPK3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412386
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB2701851
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701770
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA003995
Anti-MAPK3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005700
Anti-MAPK3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104574
Anti-MAPK3 antibody produced in rabbit, affinity isolated antibody
SAB2104732
Anti-MAPK3 antibody produced in rabbit, affinity isolated antibody
M3946 Anti-MNK1 antibody produced in rabbit, ~1.4 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503422
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4503423
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB2700025
Anti-NS5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700182
Anti-NS5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502389
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4502390
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4300194
Anti-phospho-MAPK3 (pThr202) antibody produced in rabbit, affinity isolated antibody
SAB4300195
Anti-phospho-MAPK3 (pTyr204) antibody produced in rabbit, affinity isolated antibody
SAB4504626
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
SAB4504714
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
R5773
Anti-Raf-1 (631-648) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300291
Anti-RAF1 (Ab-259) antibody produced in rabbit, affinity isolated antibody
SAB4300462
Anti-RAF1 (Ab-338) antibody produced in rabbit, affinity isolated antibody