EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
The involvement of sigma1 receptors in donepezil-induced rescue of hippocampal LTP impaired by beta-amyloid peptide.
PMID 24956443
Abstract
Donepezil is a potent acetylcholinesterase inhibitor used for the treatment of Alzheimer's disease (AD). Additional therapeutically relevant target for donepezil is sigma1 receptor (Sig1-R). Beta-amyloid peptide (Aβ) is believed to contribute to the pathogenesis of AD. In our previous work (Kapai et al., 2012), we have shown that donepezil antagonizes the suppressive action of Aβ(1-42) on long-term potentiation (LTP) in rat hippocampal slices. The purpose of the present study was to determine whether Sig1-R is involved into the mechanisms of donepezil action. For this purpose, we have tested whether agonist of Sig1-R PRE-084 mimics, and antagonist of Sig1-R haloperidol abolishes the effect of donepezil. Population spikes (PSs) were recorded from the pyramidal layer of the CA1 region of rat hippocampal slices. Drugs were applied by addition to the perfusate starting 15 min before and ending 5 min after the tetanus. In the control group, the amplitude of PS 30 min post-tetanus reached 153±10%. Aβ (200 nM) markedly suppressed the LTP magnitude or even caused the suppression of baseline PS (82±8%, P<0.001). This suppression of LTP could be markedly prevented when 1 μM donepezil was co-administered with Aβ (136±11%, P<0.05). Further, we co-administered three substances: Aβ, donepezil and 0.5 μM haloperidol and have found that haloperidol antagonized the stimulating effect of donepezil on LTP (92±6%, P<0.05). Agonist of Sig1-R PRE-084 (0.1-10 μM) enhanced control LTP and abolished the inhibitory effect of Aβ on LTP in a concentration-dependent manner. The amplitude of PS 30 min post-tetanus reached 183±7% (P<0.01) for 10 μM PRE-084. The results suggest that activation of Sig1-R is involved into the mechanisms of donepezil-induced rescue of hippocampal LTP impaired by Aβ.