EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Neuroprotection of atractylenolide III from Atractylodis macrocephalae against glutamate-induced neuronal apoptosis via inhibiting caspase signaling pathway.
PMID 24958167
Abstract
Glutamate-induced excitotoxicity appears to play a crucial role in neurological disorders. Neuroprotection against glutamate-induced excitotoxicity has been proposed as a therapeutic strategy for preventing and/or treating these excitotoxicity-mediated diseases. In the present study, atractylenolide III, which exhibited significantly neuroprotective effect against glutamate-induced neuronal apoptosis, was isolated from Atractylodes macrocephala by means of bioactivity-guided fractionation. The inhibitory effect of atractylenolide III on glutamate-induced neuronal apoptosis was in a concentration-dependent manner. The anti-apoptotic property of atractylenolide III might be mediated, in part, via inhibiting caspase signaling pathway. Atractylenolide III may have therapeutic potential in excitotoxicity-mediated neurological diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1086
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, ≥97% (HPLC), powder
C30H37N5O13