EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
Galectin-9 controls the therapeutic activity of 4-1BB-targeting antibodies.
PMID 24958847
Abstract
Biologics to TNF family receptors are prime candidates for therapy of immune disease. Whereas recent studies have highlighted a requirement for Fcγ receptors in enabling the activity of CD40, TRAILR, and GITR when engaged by antibodies, other TNFR molecules may be controlled by additional mechanisms. Antibodies to 4-1BB (CD137) are currently in clinical trials and can both augment immunity in cancer and promote regulatory T cells that inhibit autoimmune disease. We found that the action of agonist anti-4-1BB in suppressing autoimmune and allergic inflammation was completely dependent on Galectin-9 (Gal-9). Gal-9 directly bound to 4-1BB, in a site distinct from the binding site of antibodies and the natural ligand of 4-1BB, and Gal-9 facilitated 4-1BB aggregation, signaling, and functional activity in T cells, dendritic cells, and natural killer cells. Conservation of the Gal-9 interaction in humans has important implications for effective clinical targeting of 4-1BB and possibly other TNFR superfamily molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22461F Anti-Interleukin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8398 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I8273 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7401 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7784 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2501430
Anti-Lgals3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406081
Anti-LGALS3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003162
Anti-LGALS3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101338
Anti-LGALS3 antibody produced in rabbit, affinity isolated antibody
SAB1410755
Anti-LGALS3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2107361
Anti-TNFRSF4 antibody produced in rabbit, affinity isolated antibody
RAB0303
Human IL-5 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0305
Human IL-5 R α ELISA Kit, for serum, plasma, cell culture supernatant and urine
SRP3242 IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I0523 Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I4161 Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
SAB4700392 Monoclonal Anti-CD11c-FITC antibody produced in mouse, clone BU15, purified immunoglobulin, buffered aqueous solution
WH0003558M3 Monoclonal Anti-IL2 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
I6784 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5334, purified immunoglobulin, lyophilized powder
I6909 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5355, purified immunoglobulin, lyophilized powder
WH0003958M1
Monoclonal Anti-LGALS3 antibody produced in mouse, clone 3C2-2A3, purified immunoglobulin, buffered aqueous solution
RAB0304
Mouse IL-5 ELISA Kit, for serum, plasma and cell culture supernatant