EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
HTLV-1 induces a Th1-like state in CD4+CCR4+ T cells.
PMID 24960164
Abstract
Human T-lymphotropic virus type 1 (HTLV-1) is linked to multiple diseases, including the neuroinflammatory disease HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP) and adult T cell leukemia/lymphoma. Evidence suggests that HTLV-1, via the viral protein Tax, exploits CD4+ T cell plasticity and induces transcriptional changes in infected T cells that cause suppressive CD4+CD25+CCR4+ Tregs to lose expression of the transcription factor FOXP3 and produce IFN-γ, thus promoting inflammation. We hypothesized that transformation of HTLV-1-infected CCR4+ T cells into Th1-like cells plays a key role in the pathogenesis of HAM/TSP. Here, using patient cells and cell lines, we demonstrated that Tax, in cooperation with specificity protein 1 (Sp1), boosts expression of the Th1 master regulator T box transcription factor (T-bet) and consequently promotes production of IFN-γ. Evaluation of CSF and spinal cord lesions of HAM/TSP patients revealed the presence of abundant CD4+CCR4+ T cells that coexpressed the Th1 marker CXCR3 and produced T-bet and IFN-γ. Finally, treatment of isolated PBMCs and CNS cells from HAM/TSP patients with an antibody that targets CCR4+ T cells and induces cytotoxicity in these cells reduced both viral load and IFN-γ production, which suggests that targeting CCR4+ T cells may be a viable treatment option for HAM/TSP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100369
Anti-CCR4 antibody produced in rabbit, affinity isolated antibody
HPA031613
Anti-CCR4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500327
Anti-CCR4 antibody produced in rabbit, affinity isolated antibody
SAB2100457
Anti-CNOT6 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA044568
Anti-CNOT6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C1366 Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501241
Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody
SAB3501088
Anti-CXCR3 antibody produced in rabbit, affinity isolated antibody
SAB1407427
Anti-FOXP3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410887
Anti-FOXP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501136
Anti-GATA3 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405853
Anti-GATA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV32548
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody
SAB2100898
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody
SAB3500250
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA029731
Anti-GATA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501126
Anti-GATA3 antibody produced in rabbit, affinity isolated antibody
SAB1409959
Anti-SGPL1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408645
Anti-SGPL1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV37192
Anti-SP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV37193
Anti-SP1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500981
Anti-SP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412220
ANTI-SP1 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1412261
ANTI-SP1 antibody produced in mouse, clone 4B11, purified immunoglobulin, buffered aqueous solution
SAB1412262
ANTI-SP1 antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB1412263
ANTI-SP1 antibody produced in mouse, clone 3H7, purified immunoglobulin, buffered aqueous solution
SAB1412264
ANTI-SP1 antibody produced in mouse, clone 1G9, purified immunoglobulin, buffered aqueous solution
S9809 Anti-Sp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001853
Anti-SP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43585
Anti-SP1 antibody produced in rabbit, affinity isolated antibody