EMAIL THIS PAGE TO A FRIEND
ACS nano
Electrical transport and grain growth in solution-cast, chloride-terminated cadmium selenide nanocrystal thin films.
PMID 24960255
Abstract
We report the evolution of electrical transport and grain size during the sintering of thin films spin-cast from soluble phosphine and amine-bound, chloride-terminated cadmium selenide nanocrystals. Sintering of the nanocrystals occurs in three distinct stages as the annealing temperature is increased: (1) reversible desorption of the organic ligands (≤150 °C), (2) irreversible particle fusion (200-300 °C), and (3) ripening of the grains to >5 nm domains (>200 °C). Grain growth occurs at 200 °C in films with 8 atom % Cl(-), while films with 3 atom % Cl(-) resist growth until 300 °C. Fused nanocrystalline thin films (grain size = 4.5-5.5 nm) on thermally grown silicon dioxide gate dielectrics produce field-effect transistors with electron mobilities as high as 25 cm(2)/(Vs) and on/off ratios of 10(5) with less than 0.5 V hysteresis in threshold voltage without the addition of indium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175870
Benzene-d6, "100%", 99.96 atom % D
C6D6
561509
Benzene-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
C6D6
151815
Benzene-d6, 99.6 atom % D
C6D6
308722
Benzene-d6, 99.6 atom % D
C6D6
364940
Benzene-d6, 99.6 atom % D, contains 0.03 % (v/v) TMS
C6D6
175978
Benzene-d6, 99 atom % D
C6D6
570680
Benzene-d6, anhydrous, ≥99.6 atom % D
C6D6
19480
Butylamine, purum, ≥99.0% (GC)
C4H11N
W313009
Butylamine, ≥99%
C4H11N
471305
Butylamine, 99.5%
C4H11N
90893
Butylamine, analytical reference material
C4H11N
386529
Chlorotrimethylsilane, purified by redistillation, ≥99%
C3H9ClSi
95541
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C3H9ClSi
92361
Chlorotrimethylsilane, ≥98.0% (GC)
C3H9ClSi
92360
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
C3H9ClSi
89595
Chlorotrimethylsilane, for GC derivatization, ≥99.0% (GC)
C3H9ClSi
385433
Chlorotrimethylsilane solution, 1.0 M in methylene chloride
C3H9ClSi
384410
Chlorotrimethylsilane solution, 1.0 M in THF
C3H9ClSi
45999
Methyl acetate, for HPLC, ≥99.8%
C3H6O2
45997
Methyl acetate, analytical standard
C3H6O2
W267600
Methyl acetate, ≥98%, FG
C3H6O2
296996
Methyl acetate, anhydrous, 99.5%
C3H6O2
537039
Methyl acetate, reagent grade, 95%
C3H6O2
19-2340
Methyl acetate, JIS special grade, ≥99.5%
C3H6O2
186325
Methyl acetate, ReagentPlus®, 99%
C3H6O2
W267619
Methyl acetate, natural, 98%, FG
C3H6O2
T49484
Tri-n-butylphosphine, 97%
C12H27P
731374
Tri-n-butylphosphine, 99%
C12H27P
247049
Tributylphosphine, mixture of isomers, 97%
C12H27P
90827
Tributylphosphine, ≥93.5% (Tri-N-butylphosphine, GC)
C12H27P