EMAIL THIS PAGE TO A FRIEND
BMC microbiology
Perturbation of copper homeostasis is instrumental in early developmental arrest of intraerythrocytic Plasmodium falciparum.
PMID 24961242
Abstract
Malaria continues to be a devastating disease. The elucidation of factors inducing asexual growth versus arrest of Plasmodium falciparum can provide information about the development of the parasite, and may help in the search for novel malaria medication. Based on information from genome-wide transcriptome profiling of different developmental stages of P. falciparum, we investigated the critical importance of copper homeostasis in the developmental succession of P. falciparum with regard to three aspects of copper function. These were:1) inhibition of copper-binding proteins, 2) copper-ion chelation, and 3) down-regulated expression of genes encoding copper-binding proteins associated with a specific growth-promoting factor. Inhibition of copper-binding proteins with tetrathiomolybdate (TTM) caused cessation of growth of the parasite. TTM arrested the parasite irreversibly during the trophozoite to schizont stage progression. Target molecules for TTM may be present in P. falciparum. The involvement of copper ions in developmental arrest was also investigated by copper-ion chelating methods, which indicated a critical function of reduced copper ions (Cu1+) in the parasite during the early developmental stage. Copper ions, not only in the parasite but also in host cells, were targets of the chelators. Chelation of Cu1+caused blockage of trophozoite progression from the ring stage. Profound growth arrest was detected in parasites cultured in a chemically defined medium containing hexadecanoic acid alone as a growth-promoting factor. This developmental arrest was associated with down-regulated expression of genes encoding copper-binding proteins. Cis-9-octadecenoic acid completely prevented the down-regulation of gene expression and developmental arrest that were observed with the use of hexadecanoic acid. The critical importance of copper homeostasis in early developmental stages of P. falciparum was confirmed. Perturbation of copper homeostasis induced profound and early developmental arrest of P. falciparum. These findings should help to elucidate the mechanisms behind the development of P. falciparum, and may be applied in the development of effective antimalarial strategies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W302902
D-Sorbitol, ≥98%, FCC, FG
C6H14O6
240850
D-Sorbitol, 99%
C6H14O6
S1876
D-Sorbitol, ≥98%
C6H14O6
85529
D-Sorbitol, BioUltra, ≥99.5% (HPLC)
C6H14O6
S6021
D-Sorbitol, for molecular biology, ≥98%
C6H14O6
85530
D-Sorbitol, crystallized, ≥99.0% (HPLC)
C6H14O6
S7547
D-Sorbitol, BioXtra, ≥98%
C6H14O6
S3889
D-Sorbitol, BioReagent, cell culture tested, plant cell culture tested
C6H14O6
S0900
D-Sorbitol, for electrophoresis
C6H14O6
S8143
D-Sorbitol, plant cell culture tested
C6H14O6
97336
D-Sorbitol, liquid, tested according to Ph.Eur.
C6H14O6
47841
D-Sorbitol, analytical standard
C6H14O6
Y0000441
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
C5H4N4O
77662
Hypoxanthine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C5H4N4O
H9377
Hypoxanthine, ≥99.0%
C5H4N4O
H9636
Hypoxanthine, powder, BioReagent, suitable for cell culture
C5H4N4O
N1501
Neocuproine, ≥98%
C14H12N2
1617000
Sorbitol, United States Pharmacopeia (USP) Reference Standard
C6H14O6
S1000000
Sorbitol, European Pharmacopoeia (EP) Reference Standard
C6H14O6
PHR1006
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14O6
309532
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
C6H10O6