EMAIL THIS PAGE TO A FRIEND
Oncotarget
The poison oligonucleotide F10 is highly effective against acute lymphoblastic leukemia while sparing normal hematopoietic cells.
PMID 24961587
Abstract
F10 is an oligonucleotide based on the thymidylate synthase (TS) inhibitory 5-fluorouracil (5-FU) metabolite, 5-fluoro-2'-deoxyuridine-5'-O-monophosphate. We sought to determine the activity of F10 against preclinical models of acute lymphoblastic leukemia (ALL). F10 treatment resulted in robust induction of apoptosis that could not be equaled by 100 fold more 5-FU. F10 was more potent than Ara-C and doxorubicin against a panel of murine and human ALL cells with an average IC50 value of 1.48 nM (range 0.07 to 5.4 nM). F10 was more than 1000 times more potent than 5-FU. In vivo, F10 treatment significantly increased survival in 2 separate syngeneic ALL mouse models and 3 separate xenograft models. F10 also protected mice from leukemia-induced weight loss. In ALL cells made resistant to Ara-C, F10 remained highly active in vitro and in vivo. Using labeled F10, uptake by the ALL cell lines DG75 and SUP-B15 was rapid and profoundly temperature-dependent. Both cell lines demonstrated increased uptake compared to normal murine lineage- depleted marrow cells. Consistent with this decreased uptake, F10 treatment did not alter the ability of human hematopoietic stem cells to engraft in immunodeficient mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
SAB4300363
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2102511
Anti-TP53 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102512
Anti-TP53 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2102513
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2102514
Anti-TP53 (ab4) antibody produced in rabbit, affinity isolated antibody
SAB2102515
Anti-TP53 (ab5) antibody produced in rabbit, affinity isolated antibody
SAB1406544
Anti-TP53 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV02055
Anti-TP53 antibody produced in rabbit, IgG fraction of antiserum
AV37163
Anti-TRP53 antibody produced in rabbit, IgG fraction of antiserum
SAB2106418
Anti-TRP53 antibody produced in rabbit, affinity isolated antibody
P6749
Monoclonal Anti-p53 antibody produced in mouse, clone PAb1801, purified from hybridoma cell culture
AMAB90956
Monoclonal Anti-p53 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2199, purified immunoglobulin, buffered aqueous glycerol solution
P5813
Monoclonal Anti-p53 antibody produced in mouse, clone BP53-12, ascites fluid
P8999
Monoclonal Anti-p53 antibody produced in mouse, clone DO-7, purified from hybridoma cell culture
P7874 Monoclonal Anti-p53 antibody produced in mouse, ~2 mg/mL, clone DO-2, purified immunoglobulin, buffered aqueous solution
P6874
Monoclonal Anti-p53 antibody produced in mouse, clone DO-1, purified from hybridoma cell culture
WH0007157M1
Monoclonal Anti-TP53 antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1404483
Monoclonal Anti-TP53 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
PLA0072
Rabbit anti-p53 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.