EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer's disease : JAD
Environmental enrichment lessens cognitive decline in APP23 mice without affecting brain sirtuin expression.
PMID 24961946
Abstract
Environmental enrichment (EE) is a non-pharmacological intervention reported to counteract pathological signs in models of Alzheimer's disease (AD). We developed EE protocols in APP23 mice and evaluated how they influenced cognitive decline and brain amyloid-β (Aβ) burden. We also investigated the involvement of sirtuins (SIRTs) as a possible molecular mediator of EE, by assessing hippocampal and cortical mRNA and protein levels of the SIRT family members (SIRT1 to SIRT7). APP23 transgenic mice were moved to EE cages (TG-EEs) starting from 3 months of age. TG-EEs were compared to transgenic mice housed in standard cages (TG-SHs) and to wild-type littermates in the two housing conditions (WT-EEs and WT-SHs). At 7 months of age, all mice were tested for behavioral performance with Morris Water Maze (MWM) and visual novel Object Recognition Test (vORT). After a month, a group underwent biochemical analyses, while another group continued in the EE environment till 18 months of age, when Aβ plaque load was assessed. At 7 months, TG-SHs had impaired behavioral performance in MWM and vORT. In contrast, TG-EE mice had restored behavioral performance. At 8 months, EE did not affect AβPP expression or processing, Aβ40/42, pGlu-Aβ3-40/3-42, or Aβ oligomer level. The expression of two Aβ degrading enzymes (insulin degrading enzyme and neprilysin) was not modulated by EE. Brain sirtuin mRNA and protein levels were unchanged, while brain-derived neurotrophic factor expression increased after EE. Aβ deposition was attenuated in 18-month-old TG-EE mice, without apparent reduction of neuroinflammatory signs. We suggest that EE had a beneficial effect on cognitive performance and lessened long-term Aβ accumulation, but brain sirtuin expression was not modulated when cognitive impairment was restored.
Related Materials