EMAIL THIS PAGE TO A FRIEND
Cell and tissue research
The role of the p53 protein in nitrosative stress-induced apoptosis of PC12 rat pheochromocytoma cells.
PMID 24962545
Abstract
PC12 rat pheochromocytoma cells are widely used to investigate signaling pathways. The p143p53PC12 cell line expresses a Val143Ala mutant p53 protein that is less capable of binding to the p53 consensus site in DNA than its wild-type counterpart. Nitric oxide (NO), depending on its concentration, is able to activate several signal transduction pathways. We used sodium nitroprusside (SNP), an NO donor compound, to analyze NO-induced cellular stress in order to clarify the mechanism and role of nitrosative stress in pathological processes, including inflammation and cancer. SNP caused cell death when applied at a concentration of 400xa0μM, p143p53PC12 cells showing higher sensitivity than wild-type PC12 cells. The mechanisms leading to the increased SNP-sensitivity of p143p53PC12 cells were then investigated. The 400-μM SNP treatment caused stress kinase activation, phosphorylation of the eukaryotic initiation factor eIF2α and p53 protein, proteolytic activation of protein kinase R, caspase-9, and caspase-3, p53 stabilization, CHOP induction, cytochrome c release from mitochondria, and a decline in the level of the Bcl-2 protein in both cell lines. All these SNP-induced changes were more robust and/or permanent in cells with the mutant p53 protein. We thus conclude that (1) the main cause of the SNP-induced apoptosis of PC12 cells is the repression of the bcl-2 gene, evoked through p53 stabilization, stress kinase activation, and CHOP induction; (2) the higher SNP sensitivity of p143p53PC12 cells is the consequence of the stronger and earlier activation of the intrinsic apoptotic pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300524
Anti-EIF2AK2 (Ab-446) antibody produced in rabbit, affinity isolated antibody
SAB4300533
Anti-EIF2AK2 (Ab-451) antibody produced in rabbit, affinity isolated antibody
SAB1412653
ANTI-EIF2AK2 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
AV09033
Anti-EIF2AK2 antibody produced in rabbit, affinity isolated antibody
HPA019795
Anti-EIF2AK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406261
Anti-PKLR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV41699
Anti-PKLR antibody produced in rabbit, IgG fraction of antiserum
SAB1410968
Anti-PKLR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
P9993 Anti-PKR (KT-16) antibody produced in rabbit, 1-2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500326
Anti-PKR antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0005610M1
Monoclonal Anti-EIF2AK2 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
SAB1404256
Monoclonal Anti-EIF2AK2, (N-terminal) antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
WH0005313M1 Monoclonal Anti-PKLR antibody produced in mouse, clone 3B11, purified immunoglobulin, buffered aqueous solution