EMAIL THIS PAGE TO A FRIEND
BMC neuroscience
HIV-1 Tat C phosphorylates VE-cadherin complex and increases human brain microvascular endothelial cell permeability.
PMID 24965120
Abstract
Human brain microvascular endothelial cells (hBMVECs) are integral part of the blood brain barrier. Post-translational modifications of adherens junction proteins regulate the permeability of human brain microvascular endothelial cells. Pro-inflammatory signals can induce tyrosine phosphorylation of adherens junction proteins. The primary objective of this work is to provide a molecular model; how the HIV-1 Tat protein can compromise the BBB integrity and eventually lead to neurological consequences. We exposed hBMVECs to recombinant HIV-1 clade C Tat protein to study the effect of HIV-1 Tat C on permeability of hBMVECs. Trans-endothelial electrical resistance and fluorescent dye migration assay have been used to check the permeability of hBMVECs. DCFDA staining has been used for intracellular reactive oxygen species (ROS) detection. Western blotting has been used to study the expression levels and co-immunoprecipitation has been used to study the interactions among adherens junction proteins. HIV-1 Tat C protein induced NOX2 and NOX4 expression level and increased intracellular ROS level. Redox-sensitive kinase; PYK2 activation led to increased tyrosine phosphorylation of VE-cadherin and β-catenin, leading to disruption of junctional assembly. The dissociation of tyrosine phosphatases VE-PTP and SHP2 from cadherin complex resulted into increased tyrosine phosphorylation of VE-cadherin and β-catenin in HIV-1 Tat C treated hBMVECs. Unrestricted phosphorylation of junctional proteins in hBMVECs, in response to HIV-1 Tat C protein; leads to the disruption of junctional complexes and increased endothelial permeability.