EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Endogenous opioid-induced neuroplasticity of dopaminergic neurons in the ventral tegmental area influences natural and opiate reward.
PMID 24966382
Abstract
Natural reward and drugs of abuse converge on the mesolimbic pathway and activate common mechanism of neural plasticity in the nucleus accumbens. Chronic exposure to opiates induces plasticity in dopaminergic neurons of the ventral tegmental area (VTA), which regulates morphine reward tolerance. Here, we test the hypotheses that mating-induced release of endogenous opioids in the VTA causes morphological changes of VTA dopamine cells in male rats, which in-turn regulate the long-term expression of experience-induced reinforcement of sexual behavior. First, sexual experience decreased VTA dopamine soma size 1 and 7 days, but not 30 days after the last mating session. This effect was blocked with naloxone before each mating session; thus, VTA dopamine cell plasticity was dependent on action of endogenous opioids. In turn, VTA plasticity was associated with altered opiate reward, as sexually experienced males did not form conditioned place preference for 0.5 mg/kg morphine. Next, it was determined whether endogenous opioid action mediates sexual reward and memory in male rats treated with naloxone during mating experience, either systemically or intra-VTA. Naloxone did not prevent the initial experience-induced facilitation of sexual behavior over repeated mating sessions, or conditioned place preference for mating. However, naloxone treatment attenuated the longer-term expression of experience-induced facilitation of sexual behavior and neural activation in mesolimbic areas induced by mating-associated conditioned cues. Together, these data demonstrate that endogenous opioids during mating induce neural plasticity in VTA dopamine neurons that appear critical for morphine reward and long-term memory for natural reward behavior.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700857 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700856 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600006 Anti-Rabbit IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB3700951 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700973 Anti-Rabbit IgG (H+L), F(ab) fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600007 Anti-Rabbit IgG (H+L), highly cross-adsorbed, Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
B7389 Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B8895 Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700844 Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700836 Anti-Rabbit IgG F(ab′)2, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700835 Anti-Rabbit IgG F(ab′)2-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
290734
Dabco® 33-LV
C6H12N2
M2392
Meprobamate, analytical standard
C9H18N2O4
M0400000
Meprobamate, European Pharmacopoeia (EP) Reference Standard
C9H18N2O4
M-039
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C9H18N2O4
M9524
Morphine sulfate salt solution, 1.0 mg/mL in methanol, drug standard
C34H38N2O6 · H2SO4
63183
N-(3-Maleimidopropionyl)biocytin, ≥75% (HPLC)
C23H33N5O7S
656895
Nickel(II) sulfate, anhydrous, 99.99% trace metals basis
NiO4S
13635
Nickel(II) sulfate hexa-/ heptahydrate, for nickel plating, DIN 50970 H, ≥20.6% Ni and Co basis
NiO4S · xH2O