EMAIL THIS PAGE TO A FRIEND
Oncology reports
MicroRNA-1 functions as a potential tumor suppressor in osteosarcoma by targeting Med1 and Med31.
PMID 24969180
Abstract
MicroRNA-1 (miR-1) has been shown to function as a critical gene regulator in multiple types of cancers. However, the role of miR-1 in osteosarcoma has not been totally clarified. In the present study, we investigated the effects of miR-1 on osteosarcoma and the underlying mechanism. We found that miR-1 was downregulated in osteosarcoma tissues and osteosarcoma cell lines. Restoration of miR-1 significantly suppressed osteosarcoma cell proliferation by inhibiting cell cycle progression. Mediator complex subunit 1 (Med1) and 31 (Med31) were validated as targets of miR-1 in osteosarcoma by luciferase reporter assay. Downregulation of Med1 and Med31 suppressed the proliferation of osteosarcoma cells, and overexpression of Med1 and Med31 abrogated the effects of miR-1 on cell proliferation. Furthermore, both miR-1 and knockdown of Med1 or Med31 reduced the expression of met proto-oncogene (MET) and blocked the downstream signaling of MET responding to hepatocyte growth factor (HGF). Taken together, the findings of this study suggest that Med1 and Med31 serve as potential gene therapeutic targets in osteosarcoma and miR-1 may prove to be a promising agent.