EMAIL THIS PAGE TO A FRIEND
Cancer research
High-throughput time-resolved FRET reveals Akt/PKB activation as a poor prognostic marker in breast cancer.
PMID 24970478
Abstract
Dysregulation of the Akt/PKB pathway has been associated with poor prognosis in several human carcinomas. Current approaches to assess Akt activation rely on intensity-based methods, which are limited by the subjectivity of manual scoring and poor specificity. Here, we report the development of a novel assay using amplified, time-resolved Förster resonance energy transfer (FRET), which is highly specific and sensitive and can be adapted to any protein. Using this approach to analyze primary breast tissue microarrays, we quantified levels of activated pAkt at a spatial resolution that revealed molecular heterogeneity within tumors. High pAkt status assessed by amplified FRET correlated with worse disease-free survival. Our findings support the use of amplified FRET to determine pAkt status in cancer tissues as candidate biomarker for the identification of high-risk patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504015
Anti-phospho-Akt (pSer246) antibody produced in rabbit, affinity isolated antibody
SAB4504331
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4504332
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504017
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
SAB4300042
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4300043
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504749
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4503852
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
SAB4300260
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
P4112 Anti-phospho-PKB (pSer473) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P3862 Anti-phospho-PKB (pThr308) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution