EMAIL THIS PAGE TO A FRIEND
Cancer research
Heparanase cooperates with Ras to drive breast and skin tumorigenesis.
PMID 24970482
Abstract
Heparanase has been implicated in cancer but its contribution to the early stages of cancer development is uncertain. In this study, we utilized nontransformed human MCF10A mammary epithelial cells and two genetic mouse models [Hpa-transgenic (Hpa-Tg) and knockout mice] to explore heparanase function at early stages of tumor development. Heparanase overexpression resulted in significantly enlarged asymmetrical acinar structures, indicating increased cell proliferation and decreased organization. This phenotype was enhanced by coexpression of heparanase variants with a mutant H-Ras gene, which was sufficient to enable growth of invasive carcinoma in vivo. These observations were extended in vivo by comparing the response of Hpa-Tg mice to a classical two-stage 12-dimethylbenz(a)anthracene (DMBA)/12-o-tetradecanoylphorbol-13-acetate (TPA) protocol for skin carcinogenesis. Hpa-Tg mice overexpressing heparanase were far more sensitive than control mice to DMBA/TPA treatment, exhibiting a 10-fold increase in the number and size of tumor lesions. Conversely, DMBA/TPA-induced tumor formation was greatly attenuated in Hpa-KO mice lacking heparanase, pointing to a critical role of heparanase in skin tumorigenesis. In support of these observations, the heparanase inhibitor PG545 potently suppressed tumor progression in this model system. Taken together, our findings establish that heparanase exerts protumorigenic properties at early stages of tumor initiation, cooperating with Ras to dramatically promote malignant development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3254
7,12-Dimethylbenz[a]anthracene, ≥95%
C20H16
442425
7,12-Dimethylbenz[a]anthracene, analytical standard
C20H16
90872
Acetone, analytical standard
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
SAB2701868
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701827
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701828 Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701689
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701690
Anti-acta2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB1306343
ANTI-FKHR (CENTER D469) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
F2303 Anti-FKHR (FOXO1a) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500938
Anti-FKHR antibody produced in rabbit, affinity isolated antibody
SAB4500939
Anti-FKHR antibody produced in rabbit, affinity isolated antibody
SAB4300410
Anti-FOXO1 (Ab-256) antibody produced in rabbit, affinity isolated antibody
SAB4300428
Anti-FOXO1 (Ab-319) antibody produced in rabbit, affinity isolated antibody
AV100607
Anti-FOXO1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32107
Anti-FOXO1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV32976
Anti-FOXO1 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB2500417
Anti-FOXO1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001252
Anti-FOXO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500507
Anti-FOXO1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1411790
ANTI-FOXO1A antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution