EMAIL THIS PAGE TO A FRIEND
Oncotarget
SCF(FBXW7α) modulates the intra-S-phase DNA-damage checkpoint by regulating Polo like kinase-1 stability.
PMID 24970797
Abstract
The intra-S-checkpoint is essential to control cell progression through S phase under normal conditions and in response to replication stress. When DNA lesions are detected, replication fork progression is blocked allowing time for repair to avoid genomic instability and the risk of cancer. DNA replication initiates at many origins of replication in eukaryotic cells, where a series of proteins form pre-replicative complexes (pre-RCs) that are activated to become pre-initiation complexes and ensure a single round of replication in each cell cycle. PLK1 plays an important role in the regulation of DNA replication, contributing to the regulation of pre-RCs formation by phosphorylating several proteins, under both normal and stress conditions. Here we report that PLK1 is ubiquitinated and degraded by SCFFBXW7α/proteasome. Moreover, we identified a new Cdc4 phosphodegron in PLK1, conserved from yeast to humans, whose mutation prevents PLK1 destruction. We established that endogenous SCFFBXW7α degrades PLK1 in the G1 and S phases of an unperturbed cell cycle and in S phase following UV irradiation. Furthermore, we showed that FBXW7α overexpression or UV irradiation prevented the loading of proteins onto chromatin to form pre-RCs and, accordingly, reduced cell proliferation. We conclude that PLK1 degradation mediated by SCFFBXW7α modulates the intra-S-phase checkpoint.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA043536
Anti-FZR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106927
Anti-FZR1 antibody produced in rabbit, affinity isolated antibody
SAB2105778
Anti-FZR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22538F Anti-GRB2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500491
Anti-GRB2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4501290
Anti-GRB2 antibody produced in rabbit, affinity isolated antibody
AV36647
Anti-MCM2 antibody produced in rabbit, IgG fraction of antiserum
SAB2101444
Anti-MCM2 antibody produced in rabbit, affinity isolated antibody
HPA031495
Anti-MCM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031496
Anti-MCM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36652
Anti-MCM7 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36653
Anti-MCM7 (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV36667
Anti-MCM7 (AB3) antibody produced in rabbit, IgG fraction of antiserum
HPA003898
Anti-MCM7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101445
Anti-MCM7 antibody produced in rabbit, affinity isolated antibody
SAB4300338
Anti-NBN (Ab-343) antibody produced in rabbit, affinity isolated antibody
HPA001429
Anti-NBN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501958
Anti-NBN antibody produced in rabbit, affinity isolated antibody
N3162
Anti-NBS1 (Nibrin) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501956
Anti-Nibrin antibody produced in rabbit, affinity isolated antibody
SAB1406265
Anti-PLK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101829
Anti-PLK1 antibody produced in rabbit, affinity isolated antibody
HPA051638 Anti-PLK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053229 Anti-PLK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502211
Anti-PLK1 antibody produced in rabbit, affinity isolated antibody
PLA0037
Goat anti-MCM2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0042
Goat anti-MCM7 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB1402580
Monoclonal Anti-FZR1 antibody produced in mouse, clone 4C4, purified immunoglobulin, buffered aqueous solution
SAB1405091
Monoclonal Anti-FZR1, (N-terminal) antibody produced in mouse, clone 3E12, purified immunoglobulin, buffered aqueous solution
WH0002885M1
Monoclonal Anti-GRB2 antibody produced in mouse, clone 4C6-H6, ascites fluid, buffered aqueous solution